Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3153891-3154152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
UniGene
Hs.18925
Gene Chromosome Location
Chr.3: 3148490-3179717 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1642234 c.846_847delCA p.Y282fs*1 Chr.3: 3154064-3154065 on Build GRCh38
5792946 c.951+10G>A p.? Chr.3: 3153950-3153950 on Build GRCh38
4116872 c.851T>C p.V284A Chr.3: 3154060-3154060 on Build GRCh38
1692303 c.895C>T p.L299F Chr.3: 3154016-3154016 on Build GRCh38
4382150 c.920G>A p.R307Q Chr.3: 3153991-3153991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053571121 Chr.3: 3153981-3153981 on Build GRCh38
rs529742781 Chr.3: 3153931-3153931 on Build GRCh38
rs774753303 Chr.3: 3153943-3153944 on Build GRCh38
rs755466110 Chr.3: 3153934-3153934 on Build GRCh38
rs975049792 Chr.3: 3154107-3154107 on Build GRCh38
rs767790560 Chr.3: 3154108-3154110 on Build GRCh38
rs142337178 Chr.3: 3153974-3153974 on Build GRCh38
rs367952702 Chr.3: 3154079-3154079 on Build GRCh38
rs746839888 Chr.3: 3154041-3154041 on Build GRCh38
rs774735724 Chr.3: 3154109-3154109 on Build GRCh38
rs756804291 Chr.3: 3153965-3153965 on Build GRCh38
rs779833529 Chr.3: 3154029-3154029 on Build GRCh38
rs766511797 Chr.3: 3154106-3154106 on Build GRCh38
rs759777570 Chr.3: 3154064-3154065 on Build GRCh38
rs375361013 Chr.3: 3154001-3154001 on Build GRCh38
rs144666584 Chr.3: 3154060-3154060 on Build GRCh38
rs751880211 Chr.3: 3153929-3153929 on Build GRCh38
rs764378133 Chr.3: 3154038-3154038 on Build GRCh38
rs758001907 Chr.3: 3154009-3154009 on Build GRCh38
rs745521075 Chr.3: 3153988-3153988 on Build GRCh38
rs748127605 Chr.3: 3154088-3154088 on Build GRCh38
rs372411530 Chr.3: 3154103-3154103 on Build GRCh38
rs181497249 Chr.3: 3153924-3153924 on Build GRCh38
rs755304141 Chr.3: 3154062-3154062 on Build GRCh38
rs757715053 Chr.3: 3154121-3154121 on Build GRCh38
rs768167718 Chr.3: 3153946-3153946 on Build GRCh38
rs773218309 Chr.3: 3154095-3154095 on Build GRCh38
rs1025412294 Chr.3: 3154120-3154120 on Build GRCh38
rs569634680 Chr.3: 3154110-3154110 on Build GRCh38
rs549822663 Chr.3: 3153957-3153957 on Build GRCh38
rs752048403 Chr.3: 3153991-3153991 on Build GRCh38
rs952604870 Chr.3: 3154121-3154121 on Build GRCh38

View Full Product Details