Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180271339-180271598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX4
Entrez Gene ID
89884
Gene Name
LIM homeobox 4
Gene Aliases
CPHD4
UniGene
Hs.658487
Gene Chromosome Location
Chr.1: 180228372-180278982 on Build GRCh38
Transcripts
NM_033343.3 XM_011510105.2 XM_011510106.2 XM_011510108.2 XM_017002755.1 NR_037642.1

Gene Symbol
LHX4-AS1
Entrez Gene ID
100527964
Gene Name
LHX4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.496545
Gene Chromosome Location
Chr.1: 180269663-180274681 on Build GRCh38
Transcripts
NM_033343.3 XM_011510105.2 XM_011510106.2 XM_011510108.2 XM_017002755.1 NR_037642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140983 c.569T>C p.L190P Chr.1: 180271497-180271497 on Build GRCh38
6700914 c.581C>T p.T194I Chr.1: 180271509-180271509 on Build GRCh38
4884728 c.597G>T p.R199S Chr.1: 180271525-180271525 on Build GRCh38
6238595 c.537C>T p.S179S Chr.1: 180271465-180271465 on Build GRCh38
900461 c.482G>A p.R161Q Chr.1: 180271410-180271410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752232336 Chr.1: 180271483-180271483 on Build GRCh38
rs776568291 Chr.1: 180271388-180271388 on Build GRCh38
rs745677990 Chr.1: 180271514-180271514 on Build GRCh38
rs121912643 Chr.1: 180271497-180271497 on Build GRCh38
rs780186723 Chr.1: 180271528-180271528 on Build GRCh38
rs746041842 Chr.1: 180271392-180271392 on Build GRCh38
rs757269879 Chr.1: 180271507-180271507 on Build GRCh38
rs762696432 Chr.1: 180271577-180271577 on Build GRCh38
rs774427400 Chr.1: 180271554-180271554 on Build GRCh38
rs752362672 Chr.1: 180271565-180271565 on Build GRCh38
rs769927908 Chr.1: 180271395-180271395 on Build GRCh38
rs767813115 Chr.1: 180271434-180271434 on Build GRCh38
rs951405837 Chr.1: 180271447-180271447 on Build GRCh38
rs770025447 Chr.1: 180271519-180271519 on Build GRCh38
rs761737345 Chr.1: 180271551-180271551 on Build GRCh38
rs145500874 Chr.1: 180271406-180271406 on Build GRCh38
rs200427160 Chr.1: 180271570-180271570 on Build GRCh38
rs370041840 Chr.1: 180271529-180271529 on Build GRCh38
rs762260848 Chr.1: 180271557-180271557 on Build GRCh38
rs786204780 Chr.1: 180271378-180271378 on Build GRCh38
rs768656562 Chr.1: 180271541-180271541 on Build GRCh38
rs183432227 Chr.1: 180271414-180271414 on Build GRCh38
rs781082569 Chr.1: 180271513-180271513 on Build GRCh38
rs750777255 Chr.1: 180271442-180271442 on Build GRCh38
rs746791648 Chr.1: 180271367-180271367 on Build GRCh38
rs200942654 Chr.1: 180271563-180271563 on Build GRCh38
rs574390212 Chr.1: 180271466-180271466 on Build GRCh38
rs558657714 Chr.1: 180271485-180271485 on Build GRCh38
rs757908143 Chr.1: 180271484-180271484 on Build GRCh38
rs958716765 Chr.1: 180271574-180271574 on Build GRCh38
rs373498977 Chr.1: 180271561-180271561 on Build GRCh38
rs772600686 Chr.1: 180271558-180271558 on Build GRCh38
rs763636316 Chr.1: 180271578-180271578 on Build GRCh38
rs777852059 Chr.1: 180271486-180271486 on Build GRCh38
rs760996220 Chr.1: 180271562-180271562 on Build GRCh38
rs774896638 Chr.1: 180271417-180271417 on Build GRCh38
rs751472840 Chr.1: 180271498-180271498 on Build GRCh38
rs762313842 Chr.1: 180271426-180271426 on Build GRCh38
rs764841257 Chr.1: 180271470-180271470 on Build GRCh38
rs747930737 Chr.1: 180271563-180271563 on Build GRCh38
rs1009287562 Chr.1: 180271405-180271405 on Build GRCh38
rs2764449 Chr.1: 180271375-180271375 on Build GRCh38

View Full Product Details