Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82553797-82554058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB1
Entrez Gene ID
64506
Gene Name
cytoplasmic polyadenylation element binding protein 1
Gene Aliases
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1
UniGene
Hs.459132
Gene Chromosome Location
Chr.15: 82543201-82648016 on Build GRCh38
Transcripts
NM_001079533.1 NM_001079534.1 NM_001079535.1 NM_001288819.1 NM_001288820.1 NM_030594.4

SNP Detail

SNP ID Chromosome Location
rs756369401 Chr.15: 82553879-82553879 on Build GRCh38
rs771946311 Chr.15: 82554010-82554010 on Build GRCh38
rs763454983 Chr.15: 82553997-82553997 on Build GRCh38
rs765804001 Chr.15: 82553825-82553826 on Build GRCh38
rs375484108 Chr.15: 82553887-82553887 on Build GRCh38
rs767734219 Chr.15: 82553844-82553844 on Build GRCh38
rs571381432 Chr.15: 82554017-82554017 on Build GRCh38
rs774953328 Chr.15: 82554001-82554001 on Build GRCh38
rs758299678 Chr.15: 82553925-82553925 on Build GRCh38
rs1006511030 Chr.15: 82553836-82553836 on Build GRCh38
rs879162018 Chr.15: 82553835-82553835 on Build GRCh38
rs372549710 Chr.15: 82553950-82553950 on Build GRCh38
rs762411267 Chr.15: 82554011-82554011 on Build GRCh38
rs752604379 Chr.15: 82553914-82553914 on Build GRCh38
rs373498878 Chr.15: 82553872-82553872 on Build GRCh38
rs760799333 Chr.15: 82554019-82554019 on Build GRCh38
rs193041779 Chr.15: 82554024-82554024 on Build GRCh38
rs746969978 Chr.15: 82553947-82553947 on Build GRCh38
rs751661655 Chr.15: 82553833-82553833 on Build GRCh38
rs770390317 Chr.15: 82553949-82553949 on Build GRCh38
rs937004475 Chr.15: 82553819-82553819 on Build GRCh38
rs752411796 Chr.15: 82554027-82554027 on Build GRCh38
rs775717963 Chr.15: 82553963-82553963 on Build GRCh38
rs534761941 Chr.15: 82553820-82553820 on Build GRCh38
rs777735904 Chr.15: 82553946-82553946 on Build GRCh38
rs769267125 Chr.15: 82554000-82554000 on Build GRCh38
rs758139832 Chr.15: 82554035-82554035 on Build GRCh38
rs757463695 Chr.15: 82553842-82553842 on Build GRCh38
rs764767501 Chr.15: 82553828-82553828 on Build GRCh38
rs1055896684 Chr.15: 82553825-82553825 on Build GRCh38
rs375580464 Chr.15: 82553952-82553952 on Build GRCh38
rs1025442150 Chr.15: 82553948-82553948 on Build GRCh38
rs769544216 Chr.15: 82553958-82553958 on Build GRCh38
rs1019713525 Chr.15: 82553998-82553998 on Build GRCh38

View Full Product Details