Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157521100-157521325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL5
Entrez Gene ID
83416
Gene Name
Fc receptor like 5
Gene Aliases
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820
UniGene
Hs.415950
Gene Chromosome Location
Chr.1: 157513377-157552520 on Build GRCh38
Transcripts
NM_001195388.1 NM_031281.2 XM_011510030.1 XM_011510031.1 XM_011510032.1 XM_011510033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5819393 c.2348T>A p.L783Q Chr.1: 157521184-157521184 on Build GRCh38
311154 c.2395C>T p.P799S Chr.1: 157521137-157521137 on Build GRCh38
6058549 c.2427G>T p.L809L Chr.1: 157521105-157521105 on Build GRCh38
6988891 c.2279G>A p.G760E Chr.1: 157521253-157521253 on Build GRCh38
4862475 c.2280G>T p.G760G Chr.1: 157521252-157521252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747628051 Chr.1: 157521231-157521231 on Build GRCh38
rs754619528 Chr.1: 157521205-157521205 on Build GRCh38
rs765290918 Chr.1: 157521255-157521255 on Build GRCh38
rs752277017 Chr.1: 157521280-157521280 on Build GRCh38
rs749212453 Chr.1: 157521236-157521236 on Build GRCh38
rs529674464 Chr.1: 157521178-157521178 on Build GRCh38
rs774152161 Chr.1: 157521241-157521241 on Build GRCh38
rs987098695 Chr.1: 157521155-157521155 on Build GRCh38
rs778589860 Chr.1: 157521290-157521290 on Build GRCh38
rs138181956 Chr.1: 157521179-157521179 on Build GRCh38
rs773144044 Chr.1: 157521130-157521130 on Build GRCh38
rs769713737 Chr.1: 157521124-157521124 on Build GRCh38
rs753909551 Chr.1: 157521152-157521152 on Build GRCh38
rs776177266 Chr.1: 157521123-157521123 on Build GRCh38
rs750978090 Chr.1: 157521257-157521257 on Build GRCh38
rs777913458 Chr.1: 157521235-157521235 on Build GRCh38
rs142943534 Chr.1: 157521289-157521289 on Build GRCh38
rs760825945 Chr.1: 157521246-157521246 on Build GRCh38
rs780376804 Chr.1: 157521199-157521199 on Build GRCh38
rs200462307 Chr.1: 157521239-157521239 on Build GRCh38
rs780580169 Chr.1: 157521230-157521230 on Build GRCh38
rs900884937 Chr.1: 157521250-157521250 on Build GRCh38
rs755731649 Chr.1: 157521288-157521288 on Build GRCh38
rs766760827 Chr.1: 157521267-157521267 on Build GRCh38
rs771543519 Chr.1: 157521177-157521177 on Build GRCh38
rs199706458 Chr.1: 157521140-157521140 on Build GRCh38
rs142243479 Chr.1: 157521248-157521248 on Build GRCh38
rs544543420 Chr.1: 157521135-157521135 on Build GRCh38
rs552385476 Chr.1: 157521240-157521240 on Build GRCh38
rs751211019 Chr.1: 157521200-157521200 on Build GRCh38
rs201833104 Chr.1: 157521191-157521191 on Build GRCh38
rs187912514 Chr.1: 157521277-157521277 on Build GRCh38
rs74858059 Chr.1: 157521276-157521276 on Build GRCh38
rs769422410 Chr.1: 157521233-157521233 on Build GRCh38
rs932974551 Chr.1: 157521142-157521142 on Build GRCh38
rs760916744 Chr.1: 157521141-157521141 on Build GRCh38
rs763421602 Chr.1: 157521259-157521259 on Build GRCh38
rs912825413 Chr.1: 157521167-157521167 on Build GRCh38
rs762905791 Chr.1: 157521132-157521132 on Build GRCh38
rs762021689 Chr.1: 157521254-157521254 on Build GRCh38
rs761392473 Chr.1: 157521134-157521135 on Build GRCh38
rs201642663 Chr.1: 157521247-157521247 on Build GRCh38
rs770833408 Chr.1: 157521138-157521138 on Build GRCh38
rs758822245 Chr.1: 157521192-157521192 on Build GRCh38

View Full Product Details