Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149623040-149623273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNA1
Entrez Gene ID
11104
Gene Name
katanin catalytic subunit A1
Gene Aliases
-
UniGene
Hs.450175
Gene Chromosome Location
Chr.6: 149594873-149649018 on Build GRCh38
Transcripts
NM_001204076.1 NM_007044.3 XM_005266804.2 XM_005266805.3 XM_017010207.1 XM_017010208.1 XM_017010209.1 XM_017010210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3749952 c.374C>A p.S125* Chr.6: 149623230-149623230 on Build GRCh38
5897584 c.457C>T p.R153C Chr.6: 149623147-149623147 on Build GRCh38
6330516 c.413G>A p.R138H Chr.6: 149623191-149623191 on Build GRCh38
3018695 c.344G>A p.R115H Chr.6: 149623260-149623260 on Build GRCh38
1074441 c.338G>T p.R113I Chr.6: 149623266-149623266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144340192 Chr.6: 149623187-149623187 on Build GRCh38
rs753192968 Chr.6: 149623081-149623081 on Build GRCh38
rs902816497 Chr.6: 149623115-149623115 on Build GRCh38
rs769899413 Chr.6: 149623218-149623218 on Build GRCh38
rs200082954 Chr.6: 149623094-149623094 on Build GRCh38
rs749913542 Chr.6: 149623203-149623203 on Build GRCh38
rs979473018 Chr.6: 149623117-149623117 on Build GRCh38
rs141453341 Chr.6: 149623200-149623200 on Build GRCh38
rs758935014 Chr.6: 149623192-149623192 on Build GRCh38
rs1023782706 Chr.6: 149623216-149623216 on Build GRCh38
rs751349445 Chr.6: 149623186-149623186 on Build GRCh38
rs745424629 Chr.6: 149623141-149623141 on Build GRCh38
rs557222260 Chr.6: 149623155-149623155 on Build GRCh38
rs765721502 Chr.6: 149623075-149623075 on Build GRCh38
rs998923102 Chr.6: 149623176-149623176 on Build GRCh38
rs201942843 Chr.6: 149623224-149623224 on Build GRCh38
rs753074118 Chr.6: 149623190-149623190 on Build GRCh38
rs771078818 Chr.6: 149623248-149623248 on Build GRCh38
rs369384760 Chr.6: 149623146-149623146 on Build GRCh38
rs142263647 Chr.6: 149623240-149623240 on Build GRCh38
rs760877623 Chr.6: 149623164-149623164 on Build GRCh38
rs553265440 Chr.6: 149623184-149623184 on Build GRCh38
rs779559933 Chr.6: 149623214-149623214 on Build GRCh38
rs773321162 Chr.6: 149623159-149623159 on Build GRCh38
rs774028107 Chr.6: 149623152-149623152 on Build GRCh38
rs373230292 Chr.6: 149623147-149623147 on Build GRCh38
rs755578956 Chr.6: 149623208-149623208 on Build GRCh38
rs899866821 Chr.6: 149623238-149623238 on Build GRCh38
rs772014499 Chr.6: 149623236-149623236 on Build GRCh38
rs771968861 Chr.6: 149623158-149623158 on Build GRCh38
rs780752509 Chr.6: 149623137-149623137 on Build GRCh38
rs778764236 Chr.6: 149623221-149623221 on Build GRCh38
rs748881542 Chr.6: 149623215-149623215 on Build GRCh38
rs766600468 Chr.6: 149623165-149623165 on Build GRCh38
rs539786823 Chr.6: 149623169-149623169 on Build GRCh38
rs139556011 Chr.6: 149623166-149623166 on Build GRCh38
rs746967618 Chr.6: 149623243-149623243 on Build GRCh38
rs755736494 Chr.6: 149623142-149623142 on Build GRCh38

View Full Product Details