Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45525448-45525705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
UniGene
Hs.24601
Gene Chromosome Location
Chr.22: 45502839-45601134 on Build GRCh38
Transcripts
NM_001996.3 NM_006485.3 NM_006486.2 NM_006487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7310130 c.317_321+8delTGAAGGTGAGAGC p.? Chr.22: 45525674-45525686 on Build GRCh38
7367831 c.239C>A p.T80K Chr.22: 45525596-45525596 on Build GRCh38
7298242 c.209A>G p.H70R Chr.22: 45525566-45525566 on Build GRCh38
4484598 c.282C>T p.P94P Chr.22: 45525639-45525639 on Build GRCh38
7367833 c.239C>A p.T80K Chr.22: 45525596-45525596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144641263 Chr.22: 45525572-45525572 on Build GRCh38
rs778380062 Chr.22: 45525684-45525684 on Build GRCh38
rs567083399 Chr.22: 45525579-45525579 on Build GRCh38
rs368018969 Chr.22: 45525626-45525626 on Build GRCh38
rs767975618 Chr.22: 45525666-45525666 on Build GRCh38
rs552439093 Chr.22: 45525642-45525642 on Build GRCh38
rs1055571235 Chr.22: 45525540-45525540 on Build GRCh38
rs377564154 Chr.22: 45525505-45525505 on Build GRCh38
rs886044038 Chr.22: 45525625-45525625 on Build GRCh38
rs370798546 Chr.22: 45525510-45525510 on Build GRCh38
rs138539092 Chr.22: 45525661-45525661 on Build GRCh38
rs1027120417 Chr.22: 45525683-45525683 on Build GRCh38
rs547185380 Chr.22: 45525487-45525487 on Build GRCh38
rs1107180 Chr.22: 45525479-45525479 on Build GRCh38
rs776464992 Chr.22: 45525506-45525506 on Build GRCh38
rs760941976 Chr.22: 45525501-45525501 on Build GRCh38
rs937300677 Chr.22: 45525483-45525483 on Build GRCh38
rs988521136 Chr.22: 45525475-45525475 on Build GRCh38
rs765918593 Chr.22: 45525636-45525636 on Build GRCh38
rs61739196 Chr.22: 45525669-45525669 on Build GRCh38
rs773164006 Chr.22: 45525615-45525615 on Build GRCh38
rs753155232 Chr.22: 45525668-45525668 on Build GRCh38
rs867023310 Chr.22: 45525470-45525470 on Build GRCh38
rs554954339 Chr.22: 45525681-45525681 on Build GRCh38
rs199802935 Chr.22: 45525643-45525643 on Build GRCh38
rs772228974 Chr.22: 45525488-45525488 on Build GRCh38
rs375315857 Chr.22: 45525534-45525534 on Build GRCh38
rs746104674 Chr.22: 45525499-45525499 on Build GRCh38
rs532634606 Chr.22: 45525622-45525622 on Build GRCh38
rs774375731 Chr.22: 45525496-45525496 on Build GRCh38
rs761714606 Chr.22: 45525547-45525547 on Build GRCh38
rs773281950 Chr.22: 45525489-45525489 on Build GRCh38

View Full Product Details