Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14374149-14374400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA13
Entrez Gene ID
6782
Gene Name
heat shock protein family A (Hsp70) member 13
Gene Aliases
STCH
UniGene
Hs.352341
Gene Chromosome Location
Chr.21: 14371115-14383188 on Build GRCh38
Transcripts
NM_006948.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7401273 c.789G>T p.L263F Chr.21: 14374244-14374244 on Build GRCh38
1029451 c.831C>T p.F277F Chr.21: 14374202-14374202 on Build GRCh38
6256470 c.832G>A p.V278M Chr.21: 14374201-14374201 on Build GRCh38
4942773 c.815A>G p.Y272C Chr.21: 14374218-14374218 on Build GRCh38
228743 c.802T>C p.Y268H Chr.21: 14374231-14374231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749118894 Chr.21: 14374332-14374332 on Build GRCh38
rs554689175 Chr.21: 14374341-14374341 on Build GRCh38
rs760873332 Chr.21: 14374193-14374193 on Build GRCh38
rs150910143 Chr.21: 14374185-14374185 on Build GRCh38
rs944311607 Chr.21: 14374212-14374212 on Build GRCh38
rs774160994 Chr.21: 14374307-14374308 on Build GRCh38
rs765672564 Chr.21: 14374210-14374210 on Build GRCh38
rs754386567 Chr.21: 14374202-14374202 on Build GRCh38
rs768697605 Chr.21: 14374292-14374292 on Build GRCh38
rs746596777 Chr.21: 14374238-14374238 on Build GRCh38
rs757697614 Chr.21: 14374209-14374209 on Build GRCh38
rs748184499 Chr.21: 14374320-14374320 on Build GRCh38
rs200321727 Chr.21: 14374218-14374218 on Build GRCh38
rs895705292 Chr.21: 14374359-14374359 on Build GRCh38
rs754494846 Chr.21: 14374281-14374281 on Build GRCh38
rs888567618 Chr.21: 14374342-14374342 on Build GRCh38
rs930199004 Chr.21: 14374310-14374310 on Build GRCh38
rs749031273 Chr.21: 14374286-14374289 on Build GRCh38
rs767826218 Chr.21: 14374180-14374180 on Build GRCh38
rs775863683 Chr.21: 14374333-14374333 on Build GRCh38
rs202139325 Chr.21: 14374298-14374298 on Build GRCh38
rs758172341 Chr.21: 14374207-14374207 on Build GRCh38
rs539991892 Chr.21: 14374172-14374172 on Build GRCh38
rs758709914 Chr.21: 14374230-14374230 on Build GRCh38
rs778151211 Chr.21: 14374302-14374302 on Build GRCh38
rs911457374 Chr.21: 14374205-14374205 on Build GRCh38
rs779711941 Chr.21: 14374233-14374233 on Build GRCh38
rs773108468 Chr.21: 14374328-14374328 on Build GRCh38
rs534883245 Chr.21: 14374340-14374340 on Build GRCh38
rs942815130 Chr.21: 14374358-14374358 on Build GRCh38
rs566295886 Chr.21: 14374327-14374327 on Build GRCh38
rs200164114 Chr.21: 14374178-14374178 on Build GRCh38
rs1043068726 Chr.21: 14374278-14374278 on Build GRCh38
rs139339783 Chr.21: 14374201-14374201 on Build GRCh38

View Full Product Details