Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38327563-38327788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR179
Entrez Gene ID
440435
Gene Name
G protein-coupled receptor 179
Gene Aliases
CSNB1E, GPR158L, GPR158L1
UniGene
Hs.462915
Gene Chromosome Location
Chr.17: 38325610-38343810 on Build GRCh38
Transcripts
NM_001004334.3

SNP Detail

SNP ID Chromosome Location
rs769955060 Chr.17: 38327665-38327665 on Build GRCh38
rs372379550 Chr.17: 38327596-38327596 on Build GRCh38
rs752430506 Chr.17: 38327713-38327713 on Build GRCh38
rs200936863 Chr.17: 38327594-38327594 on Build GRCh38
rs766171503 Chr.17: 38327605-38327605 on Build GRCh38
rs753473962 Chr.17: 38327614-38327614 on Build GRCh38
rs200274687 Chr.17: 38327642-38327642 on Build GRCh38
rs754656963 Chr.17: 38327638-38327638 on Build GRCh38
rs775470835 Chr.17: 38327589-38327589 on Build GRCh38
rs371841025 Chr.17: 38327581-38327581 on Build GRCh38
rs368927813 Chr.17: 38327697-38327697 on Build GRCh38
rs761786774 Chr.17: 38327591-38327591 on Build GRCh38
rs202208376 Chr.17: 38327757-38327757 on Build GRCh38
rs753481286 Chr.17: 38327746-38327748 on Build GRCh38
rs555226767 Chr.17: 38327700-38327700 on Build GRCh38
rs777322102 Chr.17: 38327720-38327721 on Build GRCh38
rs749287145 Chr.17: 38327688-38327688 on Build GRCh38
rs370586289 Chr.17: 38327710-38327710 on Build GRCh38
rs377617935 Chr.17: 38327748-38327748 on Build GRCh38
rs764747534 Chr.17: 38327712-38327712 on Build GRCh38
rs749758071 Chr.17: 38327586-38327586 on Build GRCh38
rs749538684 Chr.17: 38327752-38327752 on Build GRCh38
rs757876887 Chr.17: 38327728-38327728 on Build GRCh38
rs756528824 Chr.17: 38327737-38327737 on Build GRCh38
rs768726129 Chr.17: 38327692-38327692 on Build GRCh38
rs754629252 Chr.17: 38327689-38327689 on Build GRCh38
rs781671409 Chr.17: 38327731-38327731 on Build GRCh38
rs746745923 Chr.17: 38327708-38327708 on Build GRCh38
rs531671937 Chr.17: 38327625-38327625 on Build GRCh38
rs747523702 Chr.17: 38327650-38327650 on Build GRCh38
rs185715311 Chr.17: 38327587-38327587 on Build GRCh38
rs937825308 Chr.17: 38327727-38327727 on Build GRCh38
rs752102444 Chr.17: 38327727-38327727 on Build GRCh38
rs536693144 Chr.17: 38327671-38327671 on Build GRCh38
rs535949081 Chr.17: 38327597-38327597 on Build GRCh38
rs62073368 Chr.17: 38327639-38327639 on Build GRCh38
rs763714023 Chr.17: 38327720-38327720 on Build GRCh38
rs202228440 Chr.17: 38327701-38327701 on Build GRCh38
rs757639837 Chr.17: 38327662-38327662 on Build GRCh38
rs778384540 Chr.17: 38327643-38327643 on Build GRCh38
rs755556325 Chr.17: 38327717-38327718 on Build GRCh38
rs373403814 Chr.17: 38327717-38327717 on Build GRCh38
rs144104172 Chr.17: 38327664-38327664 on Build GRCh38
rs750701300 Chr.17: 38327736-38327736 on Build GRCh38

View Full Product Details