Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23706306-23706557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-6
Entrez Gene ID
137814
Gene Name
NK2 homeobox 6
Gene Aliases
CSX2, CTHM, NKX2F, NKX4-2
UniGene
Hs.532654
Gene Chromosome Location
Chr.8: 23702451-23706598 on Build GRCh38
Transcripts
NM_001136271.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4657722 c.244G>C p.E82Q Chr.8: 23706355-23706355 on Build GRCh38
4846492 c.152G>C p.R51P Chr.8: 23706447-23706447 on Build GRCh38
7333942 c.50T>A p.L17Q Chr.8: 23706549-23706549 on Build GRCh38
3699030 c.252C>T p.D84D Chr.8: 23706347-23706347 on Build GRCh38
6244325 c.183C>A p.G61G Chr.8: 23706416-23706416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376769591 Chr.8: 23706361-23706361 on Build GRCh38
rs981465849 Chr.8: 23706343-23706343 on Build GRCh38
rs767418743 Chr.8: 23706448-23706448 on Build GRCh38
rs537530270 Chr.8: 23706516-23706516 on Build GRCh38
rs558299051 Chr.8: 23706428-23706428 on Build GRCh38
rs560537458 Chr.8: 23706335-23706335 on Build GRCh38
rs772599709 Chr.8: 23706351-23706351 on Build GRCh38
rs542348801 Chr.8: 23706328-23706328 on Build GRCh38
rs545394251 Chr.8: 23706403-23706403 on Build GRCh38
rs576577900 Chr.8: 23706478-23706478 on Build GRCh38
rs770371919 Chr.8: 23706417-23706417 on Build GRCh38
rs868711551 Chr.8: 23706339-23706339 on Build GRCh38
rs779093553 Chr.8: 23706521-23706521 on Build GRCh38
rs550727329 Chr.8: 23706449-23706449 on Build GRCh38
rs982163283 Chr.8: 23706537-23706537 on Build GRCh38
rs1009399284 Chr.8: 23706501-23706501 on Build GRCh38
rs958227632 Chr.8: 23706484-23706484 on Build GRCh38
rs929350379 Chr.8: 23706430-23706430 on Build GRCh38
rs764430720 Chr.8: 23706457-23706457 on Build GRCh38
rs904420391 Chr.8: 23706535-23706535 on Build GRCh38
rs139466287 Chr.8: 23706416-23706416 on Build GRCh38
rs1007383722 Chr.8: 23706513-23706513 on Build GRCh38
rs768601589 Chr.8: 23706380-23706380 on Build GRCh38
rs772736284 Chr.8: 23706415-23706415 on Build GRCh38
rs774277973 Chr.8: 23706418-23706418 on Build GRCh38
rs1007201729 Chr.8: 23706471-23706471 on Build GRCh38
rs1038997274 Chr.8: 23706531-23706531 on Build GRCh38
rs61732911 Chr.8: 23706395-23706395 on Build GRCh38
rs757414567 Chr.8: 23706479-23706479 on Build GRCh38
rs530501230 Chr.8: 23706325-23706325 on Build GRCh38
rs760381543 Chr.8: 23706452-23706452 on Build GRCh38
rs926866298 Chr.8: 23706378-23706378 on Build GRCh38

View Full Product Details