Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42357207-42357432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLB
Entrez Gene ID
5423
Gene Name
DNA polymerase beta
Gene Aliases
-
UniGene
Hs.654484
Gene Chromosome Location
Chr.8: 42338452-42371813 on Build GRCh38
Transcripts
NM_002690.2 XM_005273535.3 XM_005273536.3 XM_005273537.3 XM_005273538.2 XM_005273539.2 XM_005273540.4 XM_006716353.2 XM_017013583.1 XM_017013584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1663609 c.479A>G p.D160G Chr.8: 42357321-42357321 on Build GRCh38
6751882 c.499delA p.V169fs*27 Chr.8: 42357341-42357341 on Build GRCh38
750322 c.512C>T p.S171F Chr.8: 42357354-42357354 on Build GRCh38
4983515 c.485T>C p.V162A Chr.8: 42357327-42357327 on Build GRCh38
6751883 c.498_499insA p.V169fs*4 Chr.8: 42357340-42357341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370103357 Chr.8: 42357277-42357277 on Build GRCh38
rs574789412 Chr.8: 42357347-42357347 on Build GRCh38
rs773971639 Chr.8: 42357276-42357276 on Build GRCh38
rs777489093 Chr.8: 42357245-42357245 on Build GRCh38
rs749106894 Chr.8: 42357249-42357249 on Build GRCh38
rs771606223 Chr.8: 42357299-42357299 on Build GRCh38
rs534555986 Chr.8: 42357315-42357315 on Build GRCh38
rs764645372 Chr.8: 42357321-42357321 on Build GRCh38
rs759143435 Chr.8: 42357287-42357287 on Build GRCh38
rs796874811 Chr.8: 42357260-42357261 on Build GRCh38
rs978581731 Chr.8: 42357373-42357373 on Build GRCh38
rs968827674 Chr.8: 42357353-42357353 on Build GRCh38
rs761102486 Chr.8: 42357318-42357318 on Build GRCh38
rs770529994 Chr.8: 42357262-42357262 on Build GRCh38
rs757539674 Chr.8: 42357323-42357323 on Build GRCh38
rs201717776 Chr.8: 42357349-42357349 on Build GRCh38
rs767234005 Chr.8: 42357340-42357341 on Build GRCh38
rs754219754 Chr.8: 42357362-42357362 on Build GRCh38
rs553756137 Chr.8: 42357398-42357398 on Build GRCh38
rs773902489 Chr.8: 42357260-42357261 on Build GRCh38
rs373761967 Chr.8: 42357335-42357335 on Build GRCh38
rs987370857 Chr.8: 42357341-42357341 on Build GRCh38
rs1011805576 Chr.8: 42357270-42357270 on Build GRCh38

View Full Product Details