Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 170214630-170214891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO3B
Entrez Gene ID
140469
Gene Name
myosin IIIB
Gene Aliases
-
UniGene
Hs.671900
Gene Chromosome Location
Chr.2: 170178145-170655325 on Build GRCh38
Transcripts
NM_001083615.3 NM_138995.4 NR_045682.1 NR_045683.1 NR_045684.1 XM_006712299.3 XM_011510654.2 XM_011510655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3895331 c.452G>A p.R151Q Chr.2: 170214754-170214754 on Build GRCh38
3981768 c.517G>A p.V173I Chr.2: 170214819-170214819 on Build GRCh38
5881306 c.463C>A p.R155S Chr.2: 170214765-170214765 on Build GRCh38
6720398 c.460C>T p.H154Y Chr.2: 170214762-170214762 on Build GRCh38
7378550 c.490C>G p.L164V Chr.2: 170214792-170214792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771339574 Chr.2: 170214865-170214865 on Build GRCh38
rs34342425 Chr.2: 170214732-170214732 on Build GRCh38
rs753523705 Chr.2: 170214836-170214836 on Build GRCh38
rs751230785 Chr.2: 170214800-170214800 on Build GRCh38
rs762553748 Chr.2: 170214721-170214721 on Build GRCh38
rs765891466 Chr.2: 170214723-170214723 on Build GRCh38
rs772742183 Chr.2: 170214843-170214843 on Build GRCh38
rs777267370 Chr.2: 170214775-170214775 on Build GRCh38
rs752627194 Chr.2: 170214818-170214818 on Build GRCh38
rs756085146 Chr.2: 170214819-170214819 on Build GRCh38
rs762480684 Chr.2: 170214779-170214779 on Build GRCh38
rs370250141 Chr.2: 170214845-170214845 on Build GRCh38
rs763398303 Chr.2: 170214797-170214797 on Build GRCh38
rs545620222 Chr.2: 170214690-170214690 on Build GRCh38
rs764644530 Chr.2: 170214692-170214692 on Build GRCh38
rs374177997 Chr.2: 170214739-170214739 on Build GRCh38
rs1054056081 Chr.2: 170214732-170214732 on Build GRCh38
rs202200514 Chr.2: 170214803-170214803 on Build GRCh38
rs752158924 Chr.2: 170214708-170214708 on Build GRCh38
rs746068059 Chr.2: 170214760-170214760 on Build GRCh38
rs201180436 Chr.2: 170214720-170214720 on Build GRCh38
rs761220272 Chr.2: 170214687-170214687 on Build GRCh38
rs750987451 Chr.2: 170214737-170214737 on Build GRCh38
rs773498914 Chr.2: 170214792-170214792 on Build GRCh38
rs79263002 Chr.2: 170214744-170214744 on Build GRCh38
rs148119655 Chr.2: 170214699-170214699 on Build GRCh38
rs550074515 Chr.2: 170214696-170214696 on Build GRCh38
rs780343511 Chr.2: 170214766-170214766 on Build GRCh38
rs752643585 Chr.2: 170214748-170214748 on Build GRCh38
rs777285605 Chr.2: 170214694-170214694 on Build GRCh38
rs765910341 Chr.2: 170214799-170214799 on Build GRCh38
rs768874499 Chr.2: 170214769-170214769 on Build GRCh38
rs770168356 Chr.2: 170214786-170214786 on Build GRCh38
rs775468660 Chr.2: 170214686-170214686 on Build GRCh38
rs111734289 Chr.2: 170214843-170214843 on Build GRCh38
rs755890172 Chr.2: 170214753-170214753 on Build GRCh38
rs200474657 Chr.2: 170214822-170214822 on Build GRCh38
rs533036085 Chr.2: 170214738-170214738 on Build GRCh38
rs4667626 Chr.2: 170214859-170214859 on Build GRCh38
rs777495303 Chr.2: 170214754-170214754 on Build GRCh38
rs551361390 Chr.2: 170214862-170214862 on Build GRCh38
rs377640353 Chr.2: 170214765-170214765 on Build GRCh38
rs920559525 Chr.2: 170214833-170214833 on Build GRCh38
rs747294806 Chr.2: 170214844-170214844 on Build GRCh38
rs80148029 Chr.2: 170214746-170214746 on Build GRCh38
rs748643739 Chr.2: 170214855-170214855 on Build GRCh38
rs758987796 Chr.2: 170214802-170214802 on Build GRCh38
rs770151516 Chr.2: 170214857-170214857 on Build GRCh38

View Full Product Details