Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50217807-50218054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A3
Entrez Gene ID
10991
Gene Name
solute carrier family 38 member 3
Gene Aliases
G17, NAT1, SN1, SNAT3
UniGene
Hs.76460
Gene Chromosome Location
Chr.3: 50205246-50220979 on Build GRCh38
Transcripts
NM_006841.5 XM_006712954.2

SNP Detail

SNP ID Chromosome Location
rs367698483 Chr.3: 50217829-50217829 on Build GRCh38
rs982770064 Chr.3: 50217861-50217861 on Build GRCh38
rs774639048 Chr.3: 50217850-50217850 on Build GRCh38
rs372237978 Chr.3: 50217962-50217962 on Build GRCh38
rs951380427 Chr.3: 50217831-50217831 on Build GRCh38
rs776790341 Chr.3: 50217974-50217974 on Build GRCh38
rs990133215 Chr.3: 50217885-50217885 on Build GRCh38
rs371471945 Chr.3: 50217856-50217856 on Build GRCh38
rs774727394 Chr.3: 50217939-50217939 on Build GRCh38
rs766189369 Chr.3: 50218016-50218016 on Build GRCh38
rs914198137 Chr.3: 50217889-50217889 on Build GRCh38
rs773629318 Chr.3: 50217827-50217827 on Build GRCh38
rs200841708 Chr.3: 50217904-50217904 on Build GRCh38
rs778163773 Chr.3: 50217855-50217865 on Build GRCh38
rs758088378 Chr.3: 50217876-50217876 on Build GRCh38
rs776111657 Chr.3: 50217877-50217877 on Build GRCh38
rs749806429 Chr.3: 50217919-50217919 on Build GRCh38
rs587683534 Chr.3: 50218015-50218015 on Build GRCh38
rs772451600 Chr.3: 50217947-50217947 on Build GRCh38
rs775760572 Chr.3: 50217862-50217862 on Build GRCh38
rs975540378 Chr.3: 50217872-50217872 on Build GRCh38
rs761369125 Chr.3: 50217896-50217896 on Build GRCh38
rs753930853 Chr.3: 50217873-50217874 on Build GRCh38
rs771342727 Chr.3: 50217833-50217833 on Build GRCh38
rs945676460 Chr.3: 50217934-50217934 on Build GRCh38
rs587617517 Chr.3: 50217864-50217864 on Build GRCh38
rs757208301 Chr.3: 50217869-50217869 on Build GRCh38
rs368739347 Chr.3: 50217961-50217961 on Build GRCh38
rs1026936563 Chr.3: 50217828-50217828 on Build GRCh38
rs759758172 Chr.3: 50217851-50217851 on Build GRCh38
rs765131121 Chr.3: 50217870-50217870 on Build GRCh38
rs587729027 Chr.3: 50217946-50217946 on Build GRCh38
rs771361067 Chr.3: 50217926-50217926 on Build GRCh38
rs587736123 Chr.3: 50217863-50217863 on Build GRCh38
rs739431 Chr.3: 50217873-50217873 on Build GRCh38
rs746639090 Chr.3: 50217887-50217887 on Build GRCh38
rs140222807 Chr.3: 50217991-50217991 on Build GRCh38
rs751227204 Chr.3: 50218018-50218018 on Build GRCh38
rs765214693 Chr.3: 50217995-50217995 on Build GRCh38
rs780665139 Chr.3: 50217905-50217905 on Build GRCh38
rs932109775 Chr.3: 50217940-50217940 on Build GRCh38
rs754599000 Chr.3: 50218019-50218019 on Build GRCh38
rs764097413 Chr.3: 50217964-50217964 on Build GRCh38
rs750360477 Chr.3: 50218007-50218007 on Build GRCh38

View Full Product Details