Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35951300-35951533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC26A8
Entrez Gene ID
116369
Gene Name
solute carrier family 26 member 8
Gene Aliases
SPGF3, TAT1
UniGene
Hs.435836
Gene Chromosome Location
Chr.6: 35943514-36024855 on Build GRCh38
Transcripts
NM_001193476.1 NM_052961.3 NM_138718.2 XM_011514294.2 XM_017010235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1078452 c.2322C>A p.F774L Chr.6: 35951313-35951313 on Build GRCh38
3075797 c.2268A>G p.I756M Chr.6: 35951464-35951464 on Build GRCh38
7081334 c.2265G>C p.L755F Chr.6: 35951467-35951467 on Build GRCh38
3729343 c.2329G>A p.A777T Chr.6: 35951306-35951306 on Build GRCh38
247380 c.2258A>G p.N753S Chr.6: 35951474-35951474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768433191 Chr.6: 35951394-35951394 on Build GRCh38
rs1056357749 Chr.6: 35951503-35951503 on Build GRCh38
rs767269587 Chr.6: 35951479-35951479 on Build GRCh38
rs758806345 Chr.6: 35951331-35951331 on Build GRCh38
rs763471368 Chr.6: 35951427-35951427 on Build GRCh38
rs764493457 Chr.6: 35951449-35951449 on Build GRCh38
rs749118043 Chr.6: 35951375-35951375 on Build GRCh38
rs761860267 Chr.6: 35951406-35951406 on Build GRCh38
rs760888170 Chr.6: 35951398-35951398 on Build GRCh38
rs745823069 Chr.6: 35951360-35951360 on Build GRCh38
rs146819478 Chr.6: 35951339-35951339 on Build GRCh38
rs751174367 Chr.6: 35951453-35951453 on Build GRCh38
rs778733240 Chr.6: 35951507-35951507 on Build GRCh38
rs373807474 Chr.6: 35951371-35951371 on Build GRCh38
rs772845742 Chr.6: 35951496-35951496 on Build GRCh38
rs756893705 Chr.6: 35951457-35951457 on Build GRCh38
rs759512722 Chr.6: 35951420-35951420 on Build GRCh38
rs375415581 Chr.6: 35951489-35951489 on Build GRCh38
rs568126801 Chr.6: 35951478-35951478 on Build GRCh38
rs914609519 Chr.6: 35951385-35951385 on Build GRCh38
rs780590188 Chr.6: 35951467-35951467 on Build GRCh38
rs564834835 Chr.6: 35951333-35951333 on Build GRCh38
rs932111639 Chr.6: 35951364-35951364 on Build GRCh38
rs948018225 Chr.6: 35951434-35951434 on Build GRCh38
rs895261710 Chr.6: 35951430-35951430 on Build GRCh38
rs368479867 Chr.6: 35951504-35951504 on Build GRCh38
rs374245672 Chr.6: 35951337-35951337 on Build GRCh38
rs753161590 Chr.6: 35951325-35951325 on Build GRCh38
rs200830295 Chr.6: 35951350-35951350 on Build GRCh38
rs115150695 Chr.6: 35951426-35951426 on Build GRCh38
rs139218243 Chr.6: 35951424-35951424 on Build GRCh38
rs746657565 Chr.6: 35951334-35951334 on Build GRCh38
rs776727865 Chr.6: 35951419-35951419 on Build GRCh38
rs201641848 Chr.6: 35951407-35951407 on Build GRCh38
rs769718714 Chr.6: 35951369-35951369 on Build GRCh38

View Full Product Details