Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63592885-63593126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB13
Entrez Gene ID
5275
Gene Name
serpin family B member 13
Gene Aliases
HSHUR7SEQ, HUR7, PI13, headpin
UniGene
Hs.241407
Gene Chromosome Location
Chr.18: 63587206-63599199 on Build GRCh38
Transcripts
NM_001307923.1 NM_012397.3 XM_005266707.3 XM_011526029.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709146 c.472G>T p.E158* Chr.18: 63592971-63592971 on Build GRCh38
80505 c.423C>T p.A141A Chr.18: 63592922-63592922 on Build GRCh38
3891240 c.424G>A p.D142N Chr.18: 63592923-63592923 on Build GRCh38
6083450 c.434G>T p.R145L Chr.18: 63592933-63592933 on Build GRCh38
263590 c.434G>A p.R145Q Chr.18: 63592933-63592933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192146478 Chr.18: 63592970-63592970 on Build GRCh38
rs149477887 Chr.18: 63592969-63592969 on Build GRCh38
rs1022370875 Chr.18: 63592972-63592972 on Build GRCh38
rs781643016 Chr.18: 63592981-63592981 on Build GRCh38
rs756656137 Chr.18: 63592984-63592984 on Build GRCh38
rs922045735 Chr.18: 63593104-63593104 on Build GRCh38
rs757922754 Chr.18: 63593017-63593020 on Build GRCh38
rs748445984 Chr.18: 63593014-63593014 on Build GRCh38
rs200313862 Chr.18: 63592989-63592989 on Build GRCh38
rs750300299 Chr.18: 63592931-63592931 on Build GRCh38
rs773083905 Chr.18: 63593019-63593019 on Build GRCh38
rs1017982654 Chr.18: 63593064-63593064 on Build GRCh38
rs187247060 Chr.18: 63592933-63592933 on Build GRCh38
rs888734658 Chr.18: 63593038-63593038 on Build GRCh38
rs143364108 Chr.18: 63592921-63592921 on Build GRCh38
rs561191607 Chr.18: 63593074-63593074 on Build GRCh38
rs558591094 Chr.18: 63592988-63592988 on Build GRCh38
rs1006538591 Chr.18: 63593052-63593052 on Build GRCh38
rs184510430 Chr.18: 63593081-63593081 on Build GRCh38
rs757540306 Chr.18: 63592976-63592976 on Build GRCh38
rs150453343 Chr.18: 63592922-63592922 on Build GRCh38
rs760475202 Chr.18: 63593021-63593021 on Build GRCh38
rs771729419 Chr.18: 63593016-63593016 on Build GRCh38
rs139349443 Chr.18: 63592916-63592916 on Build GRCh38
rs372447873 Chr.18: 63592966-63592966 on Build GRCh38
rs774716243 Chr.18: 63593082-63593082 on Build GRCh38
rs138290904 Chr.18: 63592923-63592923 on Build GRCh38
rs774689658 Chr.18: 63592999-63592999 on Build GRCh38
rs73961767 Chr.18: 63592982-63592982 on Build GRCh38
rs368306028 Chr.18: 63592932-63592932 on Build GRCh38
rs567598575 Chr.18: 63592925-63592925 on Build GRCh38
rs994792070 Chr.18: 63592987-63592987 on Build GRCh38
rs753363525 Chr.18: 63592948-63592948 on Build GRCh38

View Full Product Details