Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 174729470-174729719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSX2
Entrez Gene ID
4488
Gene Name
msh homeobox 2
Gene Aliases
CRS2, FPP, HOX8, MSH, PFM, PFM1
UniGene
Hs.89404
Gene Chromosome Location
Chr.5: 174724495-174730899 on Build GRCh38
Transcripts
NM_002449.4 XM_017009489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3393454 c.698C>T p.A233V Chr.5: 174729477-174729477 on Build GRCh38
1215624 c.785G>A p.G262D Chr.5: 174729564-174729564 on Build GRCh38
6901473 c.720C>T p.Y240Y Chr.5: 174729499-174729499 on Build GRCh38
4390575 c.695C>T p.A232V Chr.5: 174729474-174729474 on Build GRCh38
1065943 c.731G>T p.R244I Chr.5: 174729510-174729510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055211653 Chr.5: 174729675-174729675 on Build GRCh38
rs936783798 Chr.5: 174729671-174729671 on Build GRCh38
rs550795412 Chr.5: 174729648-174729648 on Build GRCh38
rs771700671 Chr.5: 174729515-174729515 on Build GRCh38
rs756385707 Chr.5: 174729550-174729550 on Build GRCh38
rs199856192 Chr.5: 174729568-174729568 on Build GRCh38
rs528373280 Chr.5: 174729558-174729558 on Build GRCh38
rs201103544 Chr.5: 174729502-174729502 on Build GRCh38
rs763019404 Chr.5: 174729626-174729626 on Build GRCh38
rs560807345 Chr.5: 174729541-174729541 on Build GRCh38
rs371825159 Chr.5: 174729505-174729505 on Build GRCh38
rs368747209 Chr.5: 174729588-174729588 on Build GRCh38
rs372223571 Chr.5: 174729598-174729598 on Build GRCh38
rs61739543 Chr.5: 174729522-174729522 on Build GRCh38
rs368570722 Chr.5: 174729501-174729501 on Build GRCh38
rs925365300 Chr.5: 174729666-174729666 on Build GRCh38
rs775044825 Chr.5: 174729519-174729519 on Build GRCh38
rs772384677 Chr.5: 174729571-174729571 on Build GRCh38
rs768358565 Chr.5: 174729591-174729591 on Build GRCh38
rs769830511 Chr.5: 174729606-174729606 on Build GRCh38
rs374244313 Chr.5: 174729528-174729528 on Build GRCh38
rs375964205 Chr.5: 174729629-174729629 on Build GRCh38
rs542254664 Chr.5: 174729535-174729535 on Build GRCh38
rs886060426 Chr.5: 174729685-174729685 on Build GRCh38
rs776218196 Chr.5: 174729530-174729530 on Build GRCh38
rs760404926 Chr.5: 174729529-174729529 on Build GRCh38
rs3087540 Chr.5: 174729617-174729617 on Build GRCh38
rs747163618 Chr.5: 174729689-174729689 on Build GRCh38
rs780486877 Chr.5: 174729583-174729583 on Build GRCh38
rs765329097 Chr.5: 174729538-174729538 on Build GRCh38
rs532365387 Chr.5: 174729643-174729643 on Build GRCh38
rs749713244 Chr.5: 174729499-174729499 on Build GRCh38
rs767820021 Chr.5: 174729548-174729548 on Build GRCh38
rs773320207 Chr.5: 174729615-174729615 on Build GRCh38
rs61739546 Chr.5: 174729532-174729532 on Build GRCh38
rs949566351 Chr.5: 174729693-174729693 on Build GRCh38
rs369125868 Chr.5: 174729658-174729658 on Build GRCh38
rs775996170 Chr.5: 174729596-174729596 on Build GRCh38
rs750227762 Chr.5: 174729540-174729540 on Build GRCh38
rs752824647 Chr.5: 174729549-174729549 on Build GRCh38
rs757757709 Chr.5: 174729567-174729567 on Build GRCh38

View Full Product Details