Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73830714-73830981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAMBP
Entrez Gene ID
10617
Gene Name
STAM binding protein
Gene Aliases
AMSH, MICCAP
UniGene
Hs.469018
Gene Chromosome Location
Chr.2: 73828911-73873656 on Build GRCh38
Transcripts
NM_006463.4 NM_201647.2 NM_213622.2 XM_005264088.3 XM_011532483.2 XM_011532484.2 XM_011532485.2 XM_017003175.1 XM_017003176.1 XM_017003177.1 XM_017003178.1 XM_017003179.1 XM_017003180.1 XM_017003181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6542726 c.72G>T p.A24A Chr.2: 73830928-73830928 on Build GRCh38
6239403 c.17A>G p.D6G Chr.2: 73830873-73830873 on Build GRCh38
5619287 c.12T>C p.H4H Chr.2: 73830868-73830868 on Build GRCh38
5633936 c.43G>T p.V15L Chr.2: 73830899-73830899 on Build GRCh38
95281 c.79G>A p.V27M Chr.2: 73830935-73830935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759853292 Chr.2: 73830937-73830937 on Build GRCh38
rs759835016 Chr.2: 73830872-73830872 on Build GRCh38
rs569782135 Chr.2: 73830857-73830857 on Build GRCh38
rs777038194 Chr.2: 73830870-73830870 on Build GRCh38
rs547933051 Chr.2: 73830848-73830848 on Build GRCh38
rs1037379712 Chr.2: 73830880-73830880 on Build GRCh38
rs765792056 Chr.2: 73830784-73830784 on Build GRCh38
rs145850419 Chr.2: 73830956-73830956 on Build GRCh38
rs750254653 Chr.2: 73830886-73830886 on Build GRCh38
rs754594431 Chr.2: 73830837-73830838 on Build GRCh38
rs529901819 Chr.2: 73830842-73830842 on Build GRCh38
rs747524076 Chr.2: 73830912-73830912 on Build GRCh38
rs777124033 Chr.2: 73830928-73830928 on Build GRCh38
rs201102191 Chr.2: 73830813-73830813 on Build GRCh38
rs988835688 Chr.2: 73830756-73830756 on Build GRCh38
rs967797784 Chr.2: 73830801-73830801 on Build GRCh38
rs748710034 Chr.2: 73830815-73830815 on Build GRCh38
rs938105850 Chr.2: 73830800-73830800 on Build GRCh38
rs768191856 Chr.2: 73830811-73830811 on Build GRCh38
rs761813085 Chr.2: 73830884-73830884 on Build GRCh38
rs772397732 Chr.2: 73830819-73830819 on Build GRCh38
rs754609431 Chr.2: 73830896-73830896 on Build GRCh38
rs1021195773 Chr.2: 73830757-73830757 on Build GRCh38
rs778405428 Chr.2: 73830897-73830897 on Build GRCh38
rs145371363 Chr.2: 73830889-73830889 on Build GRCh38
rs552232117 Chr.2: 73830879-73830879 on Build GRCh38
rs775741034 Chr.2: 73830957-73830957 on Build GRCh38
rs767731086 Chr.2: 73830960-73830960 on Build GRCh38
rs771267587 Chr.2: 73830922-73830922 on Build GRCh38
rs375857232 Chr.2: 73830890-73830890 on Build GRCh38
rs966199529 Chr.2: 73830751-73830751 on Build GRCh38
rs916491088 Chr.2: 73830895-73830895 on Build GRCh38
rs747574012 Chr.2: 73830898-73830898 on Build GRCh38
rs767316172 Chr.2: 73830885-73830885 on Build GRCh38
rs753554812 Chr.2: 73830804-73830804 on Build GRCh38
rs919629 Chr.2: 73830868-73830868 on Build GRCh38
rs968477508 Chr.2: 73830774-73830774 on Build GRCh38
rs772659635 Chr.2: 73830907-73830907 on Build GRCh38
rs748916528 Chr.2: 73830807-73830807 on Build GRCh38
rs752768984 Chr.2: 73830883-73830883 on Build GRCh38
rs140278181 Chr.2: 73830738-73830738 on Build GRCh38
rs926598211 Chr.2: 73830785-73830785 on Build GRCh38
rs917782271 Chr.2: 73830855-73830855 on Build GRCh38
rs778197130 Chr.2: 73830908-73830908 on Build GRCh38

View Full Product Details