Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19760840-19761087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX1
Entrez Gene ID
6899
Gene Name
T-box 1
Gene Aliases
CAFS, CATCH22, CTHM, DGCR, DGS, DORV, TBX1C, TGA, VCF, VCFS
UniGene
Hs.173984
Gene Chromosome Location
Chr.22: 19756703-19783593 on Build GRCh38
Transcripts
NM_005992.1 NM_080646.1 NM_080647.1 XM_006724312.2 XM_017028925.1 XM_017028926.1 XM_017028928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5520363 c.106G>A p.D36N Chr.22: 19760976-19760976 on Build GRCh38
5520362 c.106G>A p.D36N Chr.22: 19760976-19760976 on Build GRCh38
5520361 c.106G>A p.D36N Chr.22: 19760976-19760976 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931429492 Chr.22: 19760939-19760939 on Build GRCh38
rs1009463279 Chr.22: 19761056-19761057 on Build GRCh38
rs908778553 Chr.22: 19760956-19760956 on Build GRCh38
rs892361291 Chr.22: 19761039-19761039 on Build GRCh38
rs746993005 Chr.22: 19760930-19760930 on Build GRCh38
rs975164324 Chr.22: 19761058-19761058 on Build GRCh38
rs1012965841 Chr.22: 19760991-19760991 on Build GRCh38
rs770754649 Chr.22: 19760933-19760933 on Build GRCh38
rs1044423719 Chr.22: 19760997-19760997 on Build GRCh38
rs745369694 Chr.22: 19760962-19760962 on Build GRCh38
rs965143242 Chr.22: 19760913-19760913 on Build GRCh38
rs905075455 Chr.22: 19760999-19761000 on Build GRCh38
rs886038791 Chr.22: 19761015-19761016 on Build GRCh38
rs1001101215 Chr.22: 19761000-19761005 on Build GRCh38
rs975131426 Chr.22: 19760931-19760931 on Build GRCh38
rs72646952 Chr.22: 19760945-19760945 on Build GRCh38
rs1032291296 Chr.22: 19761028-19761029 on Build GRCh38
rs912338996 Chr.22: 19760859-19760859 on Build GRCh38
rs895796220 Chr.22: 19760963-19760963 on Build GRCh38
rs940065053 Chr.22: 19760960-19760960 on Build GRCh38

View Full Product Details