Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75169250-75169475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLCO2B1
Entrez Gene ID
11309
Gene Name
solute carrier organic anion transporter family member 2B1
Gene Aliases
OATP-B, OATP2B1, OATPB, SLC21A9
UniGene
Hs.7884
Gene Chromosome Location
Chr.11: 75150987-75206400 on Build GRCh38
Transcripts
NM_001145211.2 NM_001145212.2 NM_007256.4 XM_017017157.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
358576 c.655C>T p.H219Y Chr.11: 75169379-75169379 on Build GRCh38
429857 c.556C>A p.L186M Chr.11: 75169280-75169280 on Build GRCh38
2153363 c.601G>A p.V201M Chr.11: 75169325-75169325 on Build GRCh38
6053951 c.632T>G p.I211S Chr.11: 75169356-75169356 on Build GRCh38
3810434 c.532T>A p.S178T Chr.11: 75169256-75169256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770206405 Chr.11: 75169414-75169419 on Build GRCh38
rs151272312 Chr.11: 75169393-75169393 on Build GRCh38
rs75715814 Chr.11: 75169276-75169276 on Build GRCh38
rs775827694 Chr.11: 75169445-75169445 on Build GRCh38
rs767572143 Chr.11: 75169331-75169331 on Build GRCh38
rs370297980 Chr.11: 75169380-75169380 on Build GRCh38
rs773348240 Chr.11: 75169340-75169340 on Build GRCh38
rs145304530 Chr.11: 75169301-75169301 on Build GRCh38
rs375650742 Chr.11: 75169364-75169364 on Build GRCh38
rs553595767 Chr.11: 75169303-75169303 on Build GRCh38
rs113701638 Chr.11: 75169277-75169277 on Build GRCh38
rs374600854 Chr.11: 75169358-75169358 on Build GRCh38
rs201813282 Chr.11: 75169287-75169287 on Build GRCh38
rs371064308 Chr.11: 75169330-75169330 on Build GRCh38
rs746538003 Chr.11: 75169406-75169406 on Build GRCh38
rs72559740 Chr.11: 75169368-75169368 on Build GRCh38
rs770552231 Chr.11: 75169304-75169304 on Build GRCh38
rs775889132 Chr.11: 75169305-75169305 on Build GRCh38
rs777413090 Chr.11: 75169405-75169405 on Build GRCh38
rs748650777 Chr.11: 75169446-75169446 on Build GRCh38
rs374720108 Chr.11: 75169412-75169412 on Build GRCh38
rs201849801 Chr.11: 75169280-75169280 on Build GRCh38
rs754763383 Chr.11: 75169366-75169366 on Build GRCh38
rs35199625 Chr.11: 75169325-75169325 on Build GRCh38
rs202235884 Chr.11: 75169392-75169392 on Build GRCh38
rs760922757 Chr.11: 75169348-75169348 on Build GRCh38
rs780729267 Chr.11: 75169413-75169413 on Build GRCh38
rs377450276 Chr.11: 75169324-75169324 on Build GRCh38
rs545695101 Chr.11: 75169321-75169321 on Build GRCh38
rs749623098 Chr.11: 75169420-75169420 on Build GRCh38
rs753616277 Chr.11: 75169283-75169283 on Build GRCh38
rs750647248 Chr.11: 75169278-75169278 on Build GRCh38
rs1023345633 Chr.11: 75169274-75169274 on Build GRCh38
rs949860869 Chr.11: 75169319-75169319 on Build GRCh38
rs891233725 Chr.11: 75169423-75169423 on Build GRCh38
rs75987864 Chr.11: 75169312-75169312 on Build GRCh38
rs754924730 Chr.11: 75169286-75169286 on Build GRCh38
rs758464083 Chr.11: 75169399-75169399 on Build GRCh38
rs747623736 Chr.11: 75169299-75169299 on Build GRCh38
rs893086282 Chr.11: 75169384-75169384 on Build GRCh38
rs1020352224 Chr.11: 75169404-75169404 on Build GRCh38
rs769012295 Chr.11: 75169440-75169440 on Build GRCh38
rs779894847 Chr.11: 75169281-75169281 on Build GRCh38

View Full Product Details