Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48581016-48581273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STON1
Entrez Gene ID
11037
Gene Name
stonin 1
Gene Aliases
SALF, SBLF, STN1, STNB1
UniGene
Hs.44385
Gene Chromosome Location
Chr.2: 48530169-48598515 on Build GRCh38
Transcripts
NM_001198595.1 NM_006873.3 NM_001198593.1 NM_001198594.1 NM_172311.2

Gene Symbol
STON1-GTF2A1L
Entrez Gene ID
286749
Gene Name
STON1-GTF2A1L readthrough
Gene Aliases
SALF
UniGene
Hs.732016
Gene Chromosome Location
Chr.2: 48529925-48776517 on Build GRCh38
Transcripts
NM_001198595.1 NM_006873.3 NM_001198593.1 NM_001198594.1 NM_172311.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5615102 c.477G>T p.Q159H Chr.2: 48581110-48581110 on Build GRCh38
5617135 c.582C>T p.S194S Chr.2: 48581215-48581215 on Build GRCh38
3839624 c.568G>A p.E190K Chr.2: 48581201-48581201 on Build GRCh38
6480161 c.589A>T p.T197S Chr.2: 48581222-48581222 on Build GRCh38
5862599 c.637G>A p.E213K Chr.2: 48581270-48581270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538610076 Chr.2: 48581103-48581103 on Build GRCh38
rs746337577 Chr.2: 48581061-48581061 on Build GRCh38
rs753312219 Chr.2: 48581076-48581076 on Build GRCh38
rs767361834 Chr.2: 48581047-48581047 on Build GRCh38
rs755383827 Chr.2: 48581069-48581069 on Build GRCh38
rs145086840 Chr.2: 48581170-48581170 on Build GRCh38
rs760837790 Chr.2: 48581149-48581149 on Build GRCh38
rs764501385 Chr.2: 48581176-48581176 on Build GRCh38
rs1039844908 Chr.2: 48581235-48581235 on Build GRCh38
rs773405469 Chr.2: 48581054-48581054 on Build GRCh38
rs751982032 Chr.2: 48581188-48581188 on Build GRCh38
rs115483107 Chr.2: 48581139-48581139 on Build GRCh38
rs759003869 Chr.2: 48581172-48581172 on Build GRCh38
rs778438073 Chr.2: 48581206-48581206 on Build GRCh38
rs760927759 Chr.2: 48581057-48581057 on Build GRCh38
rs778578049 Chr.2: 48581237-48581237 on Build GRCh38
rs756888788 Chr.2: 48581217-48581218 on Build GRCh38
rs773022977 Chr.2: 48581147-48581147 on Build GRCh38
rs755653313 Chr.2: 48581189-48581189 on Build GRCh38
rs771827097 Chr.2: 48581146-48581146 on Build GRCh38
rs768120126 Chr.2: 48581133-48581133 on Build GRCh38
rs765768794 Chr.2: 48581074-48581074 on Build GRCh38
rs867382386 Chr.2: 48581085-48581086 on Build GRCh38
rs999657621 Chr.2: 48581092-48581092 on Build GRCh38
rs964987847 Chr.2: 48581165-48581165 on Build GRCh38
rs777488548 Chr.2: 48581217-48581217 on Build GRCh38
rs143291656 Chr.2: 48581226-48581226 on Build GRCh38
rs145361751 Chr.2: 48581222-48581222 on Build GRCh38
rs765279487 Chr.2: 48581169-48581169 on Build GRCh38
rs542933253 Chr.2: 48581223-48581223 on Build GRCh38
rs992689557 Chr.2: 48581145-48581145 on Build GRCh38
rs928109231 Chr.2: 48581118-48581118 on Build GRCh38
rs577947043 Chr.2: 48581102-48581102 on Build GRCh38
rs376805660 Chr.2: 48581107-48581107 on Build GRCh38
rs773954396 Chr.2: 48581043-48581043 on Build GRCh38
rs778321231 Chr.2: 48581083-48581083 on Build GRCh38
rs748953901 Chr.2: 48581203-48581203 on Build GRCh38
rs753321669 Chr.2: 48581086-48581087 on Build GRCh38
rs754493055 Chr.2: 48581204-48581204 on Build GRCh38
rs923242051 Chr.2: 48581182-48581182 on Build GRCh38
rs779755824 Chr.2: 48581197-48581197 on Build GRCh38
rs191162747 Chr.2: 48581082-48581082 on Build GRCh38
rs202246718 Chr.2: 48581144-48581144 on Build GRCh38
rs748039478 Chr.2: 48581207-48581207 on Build GRCh38
rs761594168 Chr.2: 48581046-48581046 on Build GRCh38
rs753975633 Chr.2: 48581063-48581063 on Build GRCh38
rs759717012 Chr.2: 48581230-48581230 on Build GRCh38
rs201775589 Chr.2: 48581163-48581163 on Build GRCh38
rs776509523 Chr.2: 48581162-48581162 on Build GRCh38

View Full Product Details