Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35137442-35137709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLE1
Entrez Gene ID
54475
Gene Name
notchless homolog 1
Gene Aliases
Nle
UniGene
Hs.85570
Gene Chromosome Location
Chr.17: 35128755-35142315 on Build GRCh38
Transcripts
NM_001014445.2 NM_018096.4 XM_017024777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
977833 c.596G>T p.S199I Chr.17: 35137582-35137582 on Build GRCh38
6334307 c.578G>T p.R193M Chr.17: 35137600-35137600 on Build GRCh38
6726679 c.583C>A p.L195I Chr.17: 35137595-35137595 on Build GRCh38
3516176 c.543C>T p.L181L Chr.17: 35137635-35137635 on Build GRCh38
5007569 c.603G>A p.W201* Chr.17: 35137575-35137575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768525328 Chr.17: 35137597-35137597 on Build GRCh38
rs1009074576 Chr.17: 35137561-35137561 on Build GRCh38
rs747370368 Chr.17: 35137496-35137496 on Build GRCh38
rs768982141 Chr.17: 35137602-35137602 on Build GRCh38
rs781772231 Chr.17: 35137567-35137567 on Build GRCh38
rs766355409 Chr.17: 35137540-35137540 on Build GRCh38
rs574114783 Chr.17: 35137553-35137553 on Build GRCh38
rs758133303 Chr.17: 35137551-35137551 on Build GRCh38
rs956957977 Chr.17: 35137626-35137626 on Build GRCh38
rs989460984 Chr.17: 35137634-35137634 on Build GRCh38
rs941246995 Chr.17: 35137488-35137488 on Build GRCh38
rs760451454 Chr.17: 35137530-35137530 on Build GRCh38
rs554232539 Chr.17: 35137526-35137526 on Build GRCh38
rs996455964 Chr.17: 35137550-35137550 on Build GRCh38
rs759855722 Chr.17: 35137579-35137579 on Build GRCh38
rs780109376 Chr.17: 35137473-35137473 on Build GRCh38
rs144949018 Chr.17: 35137543-35137543 on Build GRCh38
rs147760459 Chr.17: 35137473-35137473 on Build GRCh38
rs533571855 Chr.17: 35137563-35137563 on Build GRCh38
rs184366452 Chr.17: 35137515-35137515 on Build GRCh38
rs769876469 Chr.17: 35137517-35137517 on Build GRCh38
rs771768674 Chr.17: 35137527-35137527 on Build GRCh38
rs995979871 Chr.17: 35137532-35137532 on Build GRCh38
rs776576907 Chr.17: 35137689-35137689 on Build GRCh38
rs775597300 Chr.17: 35137523-35137523 on Build GRCh38
rs752292980 Chr.17: 35137549-35137549 on Build GRCh38
rs774482384 Chr.17: 35137536-35137537 on Build GRCh38
rs749325743 Chr.17: 35137593-35137593 on Build GRCh38
rs938520441 Chr.17: 35137475-35137475 on Build GRCh38
rs781758551 Chr.17: 35137503-35137503 on Build GRCh38
rs760506502 Chr.17: 35137624-35137624 on Build GRCh38
rs148987843 Chr.17: 35137584-35137584 on Build GRCh38
rs771622078 Chr.17: 35137500-35137500 on Build GRCh38
rs376313441 Chr.17: 35137528-35137528 on Build GRCh38
rs368427335 Chr.17: 35137592-35137592 on Build GRCh38
rs201460671 Chr.17: 35137620-35137620 on Build GRCh38
rs147948223 Chr.17: 35137566-35137566 on Build GRCh38
rs569399812 Chr.17: 35137477-35137477 on Build GRCh38
rs552252542 Chr.17: 35137681-35137681 on Build GRCh38
rs1056937150 Chr.17: 35137498-35137498 on Build GRCh38
rs764758756 Chr.17: 35137548-35137548 on Build GRCh38
rs770676543 Chr.17: 35137683-35137683 on Build GRCh38
rs759003078 Chr.17: 35137544-35137544 on Build GRCh38
rs890874721 Chr.17: 35137604-35137604 on Build GRCh38
rs552781748 Chr.17: 35137472-35137472 on Build GRCh38
rs746327917 Chr.17: 35137570-35137570 on Build GRCh38

View Full Product Details