Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100264039-100264278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF19A
Entrez Gene ID
25897
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Gene Aliases
RNF19
UniGene
Hs.292882
Gene Chromosome Location
Chr.8: 100257059-100336218 on Build GRCh38
Transcripts
NM_001280539.1 NM_015435.4 NM_183419.3 XM_005250853.3 XM_011516956.2 XM_017013302.1 XM_017013303.1 XM_017013304.1 XM_017013305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6245832 c.1460C>A p.T487K Chr.8: 100264042-100264042 on Build GRCh38
1623431 c.1419A>C p.E473D Chr.8: 100264083-100264083 on Build GRCh38
7171672 c.1367G>A p.R456Q Chr.8: 100264135-100264135 on Build GRCh38
4644825 c.1360C>G p.L454V Chr.8: 100264142-100264142 on Build GRCh38
3924394 c.1352C>T p.P451L Chr.8: 100264150-100264150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003555145 Chr.8: 100264133-100264133 on Build GRCh38
rs199534647 Chr.8: 100264224-100264226 on Build GRCh38
rs577816049 Chr.8: 100264232-100264232 on Build GRCh38
rs753883845 Chr.8: 100264190-100264190 on Build GRCh38
rs778891386 Chr.8: 100264218-100264218 on Build GRCh38
rs540029232 Chr.8: 100264233-100264233 on Build GRCh38
rs1052398343 Chr.8: 100264158-100264158 on Build GRCh38
rs760194203 Chr.8: 100264067-100264067 on Build GRCh38
rs763440155 Chr.8: 100264090-100264090 on Build GRCh38
rs1036051461 Chr.8: 100264167-100264167 on Build GRCh38
rs765669170 Chr.8: 100264076-100264076 on Build GRCh38
rs768530277 Chr.8: 100264109-100264110 on Build GRCh38
rs757255542 Chr.8: 100264205-100264205 on Build GRCh38
rs866332139 Chr.8: 100264116-100264116 on Build GRCh38
rs751894976 Chr.8: 100264130-100264130 on Build GRCh38
rs143426825 Chr.8: 100264106-100264106 on Build GRCh38
rs773654890 Chr.8: 100264078-100264078 on Build GRCh38
rs557717353 Chr.8: 100264185-100264185 on Build GRCh38
rs12544135 Chr.8: 100264196-100264196 on Build GRCh38
rs915759554 Chr.8: 100264074-100264074 on Build GRCh38
rs752748785 Chr.8: 100264164-100264164 on Build GRCh38
rs201249163 Chr.8: 100264131-100264131 on Build GRCh38
rs1045576420 Chr.8: 100264088-100264088 on Build GRCh38
rs941897213 Chr.8: 100264155-100264155 on Build GRCh38
rs774091574 Chr.8: 100264198-100264198 on Build GRCh38
rs948564514 Chr.8: 100264086-100264086 on Build GRCh38
rs767726348 Chr.8: 100264157-100264157 on Build GRCh38

View Full Product Details