Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109119331-109119556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRHR
Entrez Gene ID
7201
Gene Name
thyrotropin releasing hormone receptor
Gene Aliases
TRH-R
UniGene
Hs.3022
Gene Chromosome Location
Chr.8: 109085745-109119785 on Build GRCh38
Transcripts
NM_003301.5 XM_011517263.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062801 c.1168G>A p.A390T Chr.8: 109119426-109119426 on Build GRCh38
5605175 c.1074G>A p.K358K Chr.8: 109119332-109119332 on Build GRCh38
3643938 c.1196G>A p.*399* Chr.8: 109119454-109119454 on Build GRCh38
3643937 c.1129C>T p.L377L Chr.8: 109119387-109119387 on Build GRCh38
1094814 c.1148C>A p.S383Y Chr.8: 109119406-109119406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996674159 Chr.8: 109119503-109119503 on Build GRCh38
rs751947635 Chr.8: 109119412-109119412 on Build GRCh38
rs908211495 Chr.8: 109119419-109119419 on Build GRCh38
rs371399354 Chr.8: 109119482-109119482 on Build GRCh38
rs962512353 Chr.8: 109119395-109119395 on Build GRCh38
rs766327349 Chr.8: 109119374-109119374 on Build GRCh38
rs757802408 Chr.8: 109119418-109119418 on Build GRCh38
rs927267448 Chr.8: 109119513-109119513 on Build GRCh38
rs753034851 Chr.8: 109119397-109119397 on Build GRCh38
rs143795263 Chr.8: 109119427-109119427 on Build GRCh38
rs765526025 Chr.8: 109119392-109119392 on Build GRCh38
rs377541177 Chr.8: 109119402-109119402 on Build GRCh38
rs773072164 Chr.8: 109119363-109119363 on Build GRCh38
rs578057541 Chr.8: 109119458-109119458 on Build GRCh38
rs778788137 Chr.8: 109119394-109119395 on Build GRCh38
rs747376517 Chr.8: 109119408-109119409 on Build GRCh38
rs770880062 Chr.8: 109119500-109119500 on Build GRCh38
rs773089705 Chr.8: 109119492-109119492 on Build GRCh38
rs535374230 Chr.8: 109119446-109119446 on Build GRCh38
rs369771461 Chr.8: 109119509-109119509 on Build GRCh38
rs772134280 Chr.8: 109119402-109119403 on Build GRCh38
rs757025345 Chr.8: 109119452-109119452 on Build GRCh38
rs758826808 Chr.8: 109119401-109119401 on Build GRCh38
rs776797228 Chr.8: 109119376-109119376 on Build GRCh38
rs981576280 Chr.8: 109119448-109119448 on Build GRCh38
rs759584415 Chr.8: 109119386-109119386 on Build GRCh38
rs771483528 Chr.8: 109119409-109119410 on Build GRCh38
rs748288316 Chr.8: 109119396-109119399 on Build GRCh38
rs891177450 Chr.8: 109119530-109119530 on Build GRCh38
rs778523868 Chr.8: 109119457-109119457 on Build GRCh38
rs147214548 Chr.8: 109119439-109119439 on Build GRCh38
rs373379319 Chr.8: 109119365-109119365 on Build GRCh38
rs939113108 Chr.8: 109119369-109119369 on Build GRCh38
rs950147509 Chr.8: 109119438-109119438 on Build GRCh38
rs780924460 Chr.8: 109119518-109119518 on Build GRCh38
rs778183911 Chr.8: 109119405-109119405 on Build GRCh38
rs777152963 Chr.8: 109119410-109119411 on Build GRCh38
rs746874889 Chr.8: 109119496-109119496 on Build GRCh38
rs5776 Chr.8: 109119407-109119407 on Build GRCh38

View Full Product Details