Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54702238-54702493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD4A
Entrez Gene ID
23034
Gene Name
sterile alpha motif domain containing 4A
Gene Aliases
SAMD4, SMAUG, SMAUG1, SMG, SMGA
UniGene
Hs.733324
Gene Chromosome Location
Chr.14: 54567612-54793315 on Build GRCh38
Transcripts
NM_001161576.2 NM_015589.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4755608 c.467G>C p.S156T Chr.14: 54702335-54702335 on Build GRCh38
4820124 c.454C>T p.R152C Chr.14: 54702322-54702322 on Build GRCh38
5740104 c.491G>A p.R164H Chr.14: 54702359-54702359 on Build GRCh38
1664034 c.464C>G p.T155S Chr.14: 54702332-54702332 on Build GRCh38
1477626 c.567G>T p.W189C Chr.14: 54702435-54702435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778130983 Chr.14: 54702334-54702334 on Build GRCh38
rs757445419 Chr.14: 54702454-54702454 on Build GRCh38
rs760281497 Chr.14: 54702272-54702272 on Build GRCh38
rs1018803699 Chr.14: 54702441-54702441 on Build GRCh38
rs751896726 Chr.14: 54702426-54702426 on Build GRCh38
rs775007435 Chr.14: 54702374-54702374 on Build GRCh38
rs766714451 Chr.14: 54702413-54702413 on Build GRCh38
rs758756707 Chr.14: 54702288-54702288 on Build GRCh38
rs1050807511 Chr.14: 54702379-54702379 on Build GRCh38
rs897501571 Chr.14: 54702336-54702336 on Build GRCh38
rs767927600 Chr.14: 54702437-54702437 on Build GRCh38
rs921008978 Chr.14: 54702372-54702372 on Build GRCh38
rs776804841 Chr.14: 54702367-54702367 on Build GRCh38
rs370570421 Chr.14: 54702321-54702321 on Build GRCh38
rs1056726659 Chr.14: 54702329-54702329 on Build GRCh38
rs774885552 Chr.14: 54702267-54702267 on Build GRCh38
rs766691758 Chr.14: 54702308-54702308 on Build GRCh38
rs753545766 Chr.14: 54702284-54702284 on Build GRCh38
rs776233031 Chr.14: 54702395-54702395 on Build GRCh38
rs760016639 Chr.14: 54702428-54702428 on Build GRCh38
rs899446191 Chr.14: 54702430-54702430 on Build GRCh38
rs142168866 Chr.14: 54702332-54702333 on Build GRCh38
rs753318265 Chr.14: 54702442-54702442 on Build GRCh38
rs75516264 Chr.14: 54702336-54702337 on Build GRCh38
rs754093338 Chr.14: 54702447-54702447 on Build GRCh38
rs761499865 Chr.14: 54702405-54702405 on Build GRCh38
rs763750108 Chr.14: 54702273-54702273 on Build GRCh38
rs758334897 Chr.14: 54702441-54702442 on Build GRCh38
rs935820073 Chr.14: 54702345-54702345 on Build GRCh38
rs886738178 Chr.14: 54702421-54702421 on Build GRCh38
rs146555964 Chr.14: 54702393-54702393 on Build GRCh38
rs977311795 Chr.14: 54702299-54702299 on Build GRCh38
rs760163359 Chr.14: 54702392-54702392 on Build GRCh38
rs749591151 Chr.14: 54702358-54702358 on Build GRCh38
rs745770291 Chr.14: 54702363-54702363 on Build GRCh38
rs200776549 Chr.14: 54702353-54702353 on Build GRCh38
rs752081245 Chr.14: 54702313-54702313 on Build GRCh38
rs149215072 Chr.14: 54702445-54702445 on Build GRCh38
rs748177073 Chr.14: 54702327-54702327 on Build GRCh38
rs144215410 Chr.14: 54702359-54702359 on Build GRCh38
rs913859383 Chr.14: 54702474-54702474 on Build GRCh38
rs771934176 Chr.14: 54702370-54702370 on Build GRCh38
rs145371662 Chr.14: 54702467-54702467 on Build GRCh38
rs143416209 Chr.14: 54702446-54702446 on Build GRCh38
rs756350600 Chr.14: 54702330-54702330 on Build GRCh38
rs781289174 Chr.14: 54702322-54702322 on Build GRCh38

View Full Product Details