Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61489579-61489840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R32
Entrez Gene ID
220004
Gene Name
protein phosphatase 1 regulatory subunit 32
Gene Aliases
C11orf66, IIIG9
UniGene
Hs.729003
Gene Chromosome Location
Chr.11: 61481113-61490928 on Build GRCh38
Transcripts
NM_001170753.1 NM_145017.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4792753 c.1080A>C p.E360D Chr.11: 61489818-61489818 on Build GRCh38
5999730 c.1091G>C p.R364P Chr.11: 61489829-61489829 on Build GRCh38
6355405 c.1093G>A p.G365S Chr.11: 61489831-61489831 on Build GRCh38
7328153 c.1097G>A p.G366D Chr.11: 61489835-61489835 on Build GRCh38
4034611 c.1076A>T p.Q359L Chr.11: 61489814-61489814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771684232 Chr.11: 61489713-61489713 on Build GRCh38
rs980249744 Chr.11: 61489628-61489628 on Build GRCh38
rs751845789 Chr.11: 61489671-61489671 on Build GRCh38
rs774079964 Chr.11: 61489706-61489706 on Build GRCh38
rs141294373 Chr.11: 61489811-61489811 on Build GRCh38
rs758686796 Chr.11: 61489666-61489666 on Build GRCh38
rs781460544 Chr.11: 61489681-61489681 on Build GRCh38
rs773597031 Chr.11: 61489739-61489739 on Build GRCh38
rs565443221 Chr.11: 61489605-61489605 on Build GRCh38
rs755298458 Chr.11: 61489758-61489758 on Build GRCh38
rs752888468 Chr.11: 61489797-61489797 on Build GRCh38
rs548525691 Chr.11: 61489695-61489695 on Build GRCh38
rs79772065 Chr.11: 61489656-61489656 on Build GRCh38
rs922462525 Chr.11: 61489699-61489699 on Build GRCh38
rs777566699 Chr.11: 61489697-61489697 on Build GRCh38
rs377676964 Chr.11: 61489781-61489781 on Build GRCh38
rs765840378 Chr.11: 61489729-61489729 on Build GRCh38
rs375381435 Chr.11: 61489750-61489751 on Build GRCh38
rs760093381 Chr.11: 61489728-61489728 on Build GRCh38
rs963655049 Chr.11: 61489606-61489606 on Build GRCh38
rs142192850 Chr.11: 61489813-61489813 on Build GRCh38
rs745554615 Chr.11: 61489710-61489710 on Build GRCh38
rs374090173 Chr.11: 61489766-61489766 on Build GRCh38
rs566605530 Chr.11: 61489661-61489661 on Build GRCh38
rs1046844888 Chr.11: 61489707-61489707 on Build GRCh38
rs945074437 Chr.11: 61489704-61489704 on Build GRCh38
rs372636422 Chr.11: 61489740-61489740 on Build GRCh38
rs115927646 Chr.11: 61489755-61489755 on Build GRCh38
rs775033961 Chr.11: 61489716-61489716 on Build GRCh38
rs527905770 Chr.11: 61489642-61489642 on Build GRCh38
rs1038833477 Chr.11: 61489765-61489765 on Build GRCh38
rs1024187117 Chr.11: 61489816-61489816 on Build GRCh38
rs933700765 Chr.11: 61489702-61489702 on Build GRCh38
rs899397897 Chr.11: 61489773-61489773 on Build GRCh38
rs780499846 Chr.11: 61489670-61489670 on Build GRCh38
rs748214490 Chr.11: 61489694-61489694 on Build GRCh38
rs535105122 Chr.11: 61489680-61489680 on Build GRCh38
rs749178372 Chr.11: 61489698-61489698 on Build GRCh38
rs765654035 Chr.11: 61489796-61489797 on Build GRCh38
rs762136819 Chr.11: 61489626-61489626 on Build GRCh38
rs756149277 Chr.11: 61489800-61489800 on Build GRCh38
rs984087628 Chr.11: 61489598-61489598 on Build GRCh38

View Full Product Details