Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71537676-71537927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGT
Entrez Gene ID
8473
Gene Name
O-linked N-acetylglucosamine (GlcNAc) transferase
Gene Aliases
HINCUT-1, HRNT1, O-GLCNAC
UniGene
Hs.405410
Gene Chromosome Location
Chr.X: 71533062-71575897 on Build GRCh38
Transcripts
NM_181672.2 NM_181673.2 XM_017029907.1 XM_017029908.1

SNP Detail

SNP ID Chromosome Location
rs968357843 Chr.X: 71537773-71537773 on Build GRCh38
rs751626296 Chr.X: 71537786-71537786 on Build GRCh38
rs921044587 Chr.X: 71537770-71537770 on Build GRCh38
rs373841859 Chr.X: 71537790-71537790 on Build GRCh38
rs772555315 Chr.X: 71537791-71537792 on Build GRCh38
rs754180377 Chr.X: 71537844-71537844 on Build GRCh38
rs748448950 Chr.X: 71537790-71537791 on Build GRCh38
rs2272780 Chr.X: 71537817-71537817 on Build GRCh38
rs779514106 Chr.X: 71537886-71537886 on Build GRCh38
rs975152976 Chr.X: 71537739-71537739 on Build GRCh38
rs780294053 Chr.X: 71537841-71537841 on Build GRCh38
rs755471023 Chr.X: 71537869-71537869 on Build GRCh38
rs866232016 Chr.X: 71537744-71537744 on Build GRCh38
rs756325672 Chr.X: 71537818-71537818 on Build GRCh38
rs750704486 Chr.X: 71537804-71537804 on Build GRCh38
rs767563741 Chr.X: 71537797-71537797 on Build GRCh38
rs764047457 Chr.X: 71537784-71537784 on Build GRCh38
rs1021121430 Chr.X: 71537750-71537750 on Build GRCh38

View Full Product Details