Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23754040-23754281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCEB3
Entrez Gene ID
6924
Gene Name
transcription elongation factor B subunit 3
Gene Aliases
EloA, SIII, SIII p110, TCEB3A
UniGene
Hs.15535
Gene Chromosome Location
Chr.1: 23743366-23762059 on Build GRCh38
Transcripts
NM_003198.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5800963 c.1540_1544delTTCTC p.S515fs*31 Chr.1: 23754102-23754106 on Build GRCh38
6798161 c.1595C>A p.S532Y Chr.1: 23754157-23754157 on Build GRCh38
1127086 c.1565A>C p.E522A Chr.1: 23754127-23754127 on Build GRCh38
5800962 c.1618_1622delTTCTC p.S541fs*31 Chr.1: 23754102-23754106 on Build GRCh38
3486024 c.1635C>T p.L545L Chr.1: 23754197-23754197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188880028 Chr.1: 23754101-23754101 on Build GRCh38
rs878971041 Chr.1: 23754247-23754247 on Build GRCh38
rs779681374 Chr.1: 23754158-23754158 on Build GRCh38
rs766741607 Chr.1: 23754096-23754096 on Build GRCh38
rs1016234266 Chr.1: 23754204-23754204 on Build GRCh38
rs868073113 Chr.1: 23754079-23754079 on Build GRCh38
rs774422576 Chr.1: 23754235-23754235 on Build GRCh38
rs926917184 Chr.1: 23754236-23754236 on Build GRCh38
rs374783663 Chr.1: 23754249-23754249 on Build GRCh38
rs148243938 Chr.1: 23754253-23754253 on Build GRCh38
rs764702892 Chr.1: 23754144-23754144 on Build GRCh38
rs143293527 Chr.1: 23754147-23754147 on Build GRCh38
rs1003147141 Chr.1: 23754199-23754199 on Build GRCh38
rs771219804 Chr.1: 23754232-23754232 on Build GRCh38
rs753715107 Chr.1: 23754115-23754115 on Build GRCh38
rs776537116 Chr.1: 23754251-23754251 on Build GRCh38
rs746688714 Chr.1: 23754117-23754119 on Build GRCh38
rs368056391 Chr.1: 23754193-23754193 on Build GRCh38
rs759869258 Chr.1: 23754245-23754245 on Build GRCh38
rs747336688 Chr.1: 23754182-23754182 on Build GRCh38
rs763219140 Chr.1: 23754082-23754082 on Build GRCh38
rs749954868 Chr.1: 23754145-23754145 on Build GRCh38
rs749384778 Chr.1: 23754231-23754231 on Build GRCh38
rs371521263 Chr.1: 23754224-23754224 on Build GRCh38
rs775358723 Chr.1: 23754100-23754100 on Build GRCh38
rs915291244 Chr.1: 23754216-23754216 on Build GRCh38
rs781653623 Chr.1: 23754192-23754192 on Build GRCh38
rs201928604 Chr.1: 23754252-23754252 on Build GRCh38
rs894856215 Chr.1: 23754085-23754085 on Build GRCh38
rs553236953 Chr.1: 23754111-23754111 on Build GRCh38
rs144826294 Chr.1: 23754127-23754127 on Build GRCh38
rs764398123 Chr.1: 23754258-23754258 on Build GRCh38

View Full Product Details