Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 58527136-58527383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OMA1
Entrez Gene ID
115209
Gene Name
OMA1 zinc metallopeptidase
Gene Aliases
2010001O09Rik, DAB1, MPRP-1, YKR087C, ZMPOMA1, peptidase
UniGene
Hs.425769
Gene Chromosome Location
Chr.1: 58480719-58546799 on Build GRCh38
Transcripts
NM_145243.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6460583 c.1169A>T p.K390I Chr.1: 58527307-58527307 on Build GRCh38
3667564 c.1168A>G p.K390E Chr.1: 58527308-58527308 on Build GRCh38
1218444 c.1142A>C p.Y381S Chr.1: 58527334-58527334 on Build GRCh38
5669888 c.1200A>G p.L400L Chr.1: 58527276-58527276 on Build GRCh38
6460582 c.1169A>T p.K390I Chr.1: 58527307-58527307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748310578 Chr.1: 58527297-58527297 on Build GRCh38
rs919042013 Chr.1: 58527307-58527307 on Build GRCh38
rs770079506 Chr.1: 58527347-58527347 on Build GRCh38
rs761239007 Chr.1: 58527228-58527228 on Build GRCh38
rs745604810 Chr.1: 58527230-58527230 on Build GRCh38
rs777984806 Chr.1: 58527305-58527305 on Build GRCh38
rs771255590 Chr.1: 58527229-58527229 on Build GRCh38
rs187266868 Chr.1: 58527285-58527285 on Build GRCh38
rs750388181 Chr.1: 58527276-58527276 on Build GRCh38
rs758035870 Chr.1: 58527256-58527257 on Build GRCh38
rs376167645 Chr.1: 58527273-58527273 on Build GRCh38
rs761318149 Chr.1: 58527270-58527270 on Build GRCh38
rs753694092 Chr.1: 58527292-58527292 on Build GRCh38
rs759959079 Chr.1: 58527337-58527337 on Build GRCh38
rs777030623 Chr.1: 58527233-58527233 on Build GRCh38
rs375972697 Chr.1: 58527223-58527223 on Build GRCh38
rs776954304 Chr.1: 58527335-58527335 on Build GRCh38
rs544162374 Chr.1: 58527200-58527200 on Build GRCh38
rs763760555 Chr.1: 58527241-58527241 on Build GRCh38
rs747518874 Chr.1: 58527317-58527317 on Build GRCh38
rs771437585 Chr.1: 58527329-58527329 on Build GRCh38
rs779854559 Chr.1: 58527217-58527217 on Build GRCh38
rs779288032 Chr.1: 58527296-58527296 on Build GRCh38
rs751156811 Chr.1: 58527257-58527257 on Build GRCh38
rs759781690 Chr.1: 58527239-58527239 on Build GRCh38
rs772217197 Chr.1: 58527221-58527221 on Build GRCh38
rs768888145 Chr.1: 58527210-58527210 on Build GRCh38
rs953631738 Chr.1: 58527206-58527206 on Build GRCh38
rs145615055 Chr.1: 58527298-58527298 on Build GRCh38
rs372269511 Chr.1: 58527216-58527216 on Build GRCh38

View Full Product Details