Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114299540-114299783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGIT
Entrez Gene ID
201633
Gene Name
T-cell immunoreceptor with Ig and ITIM domains
Gene Aliases
VSIG9, VSTM3, WUCAM
UniGene
Hs.421750
Gene Chromosome Location
Chr.3: 114293986-114310288 on Build GRCh38
Transcripts
NM_173799.3 XM_011512538.1 XM_017005865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318912 c.454G>T p.V152F Chr.3: 114299659-114299659 on Build GRCh38
40107 c.441C>T p.A147A Chr.3: 114299646-114299646 on Build GRCh38
354328 c.394G>T p.A132S Chr.3: 114299599-114299599 on Build GRCh38
445339 c.394G>C p.A132P Chr.3: 114299599-114299599 on Build GRCh38
5758142 c.417G>A p.Q139Q Chr.3: 114299622-114299622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746263083 Chr.3: 114299708-114299708 on Build GRCh38
rs778828718 Chr.3: 114299607-114299607 on Build GRCh38
rs143771376 Chr.3: 114299680-114299680 on Build GRCh38
rs775210246 Chr.3: 114299715-114299715 on Build GRCh38
rs757120600 Chr.3: 114299587-114299587 on Build GRCh38
rs914483001 Chr.3: 114299679-114299679 on Build GRCh38
rs906367559 Chr.3: 114299711-114299711 on Build GRCh38
rs767613673 Chr.3: 114299740-114299740 on Build GRCh38
rs375713492 Chr.3: 114299724-114299724 on Build GRCh38
rs769049854 Chr.3: 114299620-114299642 on Build GRCh38
rs571874096 Chr.3: 114299745-114299745 on Build GRCh38
rs776955721 Chr.3: 114299703-114299703 on Build GRCh38
rs868139905 Chr.3: 114299605-114299605 on Build GRCh38
rs369133784 Chr.3: 114299647-114299647 on Build GRCh38
rs199670726 Chr.3: 114299726-114299726 on Build GRCh38
rs973073725 Chr.3: 114299678-114299678 on Build GRCh38
rs759155389 Chr.3: 114299645-114299645 on Build GRCh38
rs777259891 Chr.3: 114299682-114299682 on Build GRCh38
rs201882472 Chr.3: 114299591-114299591 on Build GRCh38
rs749509354 Chr.3: 114299593-114299593 on Build GRCh38
rs764985530 Chr.3: 114299646-114299646 on Build GRCh38
rs778198855 Chr.3: 114299690-114299690 on Build GRCh38
rs762554753 Chr.3: 114299648-114299648 on Build GRCh38
rs200224023 Chr.3: 114299709-114299709 on Build GRCh38
rs993839338 Chr.3: 114299583-114299583 on Build GRCh38
rs148148852 Chr.3: 114299696-114299696 on Build GRCh38
rs147774909 Chr.3: 114299608-114299608 on Build GRCh38
rs777787938 Chr.3: 114299568-114299568 on Build GRCh38
rs1055783473 Chr.3: 114299757-114299757 on Build GRCh38
rs201218843 Chr.3: 114299639-114299639 on Build GRCh38
rs893751401 Chr.3: 114299727-114299727 on Build GRCh38
rs372970765 Chr.3: 114299688-114299688 on Build GRCh38
rs373179347 Chr.3: 114299725-114299725 on Build GRCh38
rs141092303 Chr.3: 114299651-114299651 on Build GRCh38
rs73238032 Chr.3: 114299594-114299594 on Build GRCh38
rs966813707 Chr.3: 114299602-114299602 on Build GRCh38
rs763870601 Chr.3: 114299649-114299649 on Build GRCh38
rs201631374 Chr.3: 114299746-114299746 on Build GRCh38
rs751360402 Chr.3: 114299577-114299577 on Build GRCh38
rs147577671 Chr.3: 114299689-114299689 on Build GRCh38
rs779221087 Chr.3: 114299683-114299683 on Build GRCh38
rs750285192 Chr.3: 114299676-114299676 on Build GRCh38
rs767417384 Chr.3: 114299662-114299662 on Build GRCh38
rs375103800 Chr.3: 114299620-114299620 on Build GRCh38
rs146957667 Chr.3: 114299652-114299652 on Build GRCh38
rs149263615 Chr.3: 114299622-114299622 on Build GRCh38
rs962636157 Chr.3: 114299562-114299562 on Build GRCh38
rs141793043 Chr.3: 114299691-114299691 on Build GRCh38

View Full Product Details