Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161210143-161210372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFS2
Entrez Gene ID
4720
Gene Name
NADH:ubiquinone oxidoreductase core subunit S2
Gene Aliases
CI-49
UniGene
Hs.173611
Gene Chromosome Location
Chr.1: 161197377-161214395 on Build GRCh38
Transcripts
NM_001166159.1 NM_004550.4 XM_005245208.2 XM_005245209.1 XM_017001357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
245983 c.805C>T p.R269* Chr.1: 161210328-161210328 on Build GRCh38
5546984 c.739delT p.S248fs*15 Chr.1: 161210147-161210147 on Build GRCh38
1159812 c.737A>C p.Q246P Chr.1: 161210145-161210145 on Build GRCh38
2086995 c.760C>T p.R254W Chr.1: 161210168-161210168 on Build GRCh38
211966 c.766G>C p.D256H Chr.1: 161210174-161210174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760243514 Chr.1: 161210275-161210275 on Build GRCh38
rs751955645 Chr.1: 161210329-161210329 on Build GRCh38
rs368115251 Chr.1: 161210264-161210264 on Build GRCh38
rs981193227 Chr.1: 161210236-161210236 on Build GRCh38
rs752147633 Chr.1: 161210209-161210209 on Build GRCh38
rs755515997 Chr.1: 161210212-161210212 on Build GRCh38
rs200452962 Chr.1: 161210223-161210223 on Build GRCh38
rs755330962 Chr.1: 161210339-161210339 on Build GRCh38
rs766896258 Chr.1: 161210317-161210317 on Build GRCh38
rs763804955 Chr.1: 161210185-161210185 on Build GRCh38
rs912874455 Chr.1: 161210334-161210334 on Build GRCh38
rs772040956 Chr.1: 161210270-161210270 on Build GRCh38
rs987556298 Chr.1: 161210298-161210298 on Build GRCh38
rs761521965 Chr.1: 161210300-161210300 on Build GRCh38
rs115682162 Chr.1: 161210271-161210271 on Build GRCh38
rs759529259 Chr.1: 161210267-161210267 on Build GRCh38
rs778081445 Chr.1: 161210231-161210231 on Build GRCh38
rs758853352 Chr.1: 161210199-161210199 on Build GRCh38
rs371923433 Chr.1: 161210261-161210261 on Build GRCh38
rs763470896 Chr.1: 161210288-161210288 on Build GRCh38
rs111353446 Chr.1: 161210204-161210204 on Build GRCh38
rs753555709 Chr.1: 161210294-161210294 on Build GRCh38
rs374901170 Chr.1: 161210262-161210262 on Build GRCh38
rs199529604 Chr.1: 161210238-161210238 on Build GRCh38
rs781199811 Chr.1: 161210220-161210220 on Build GRCh38
rs536763398 Chr.1: 161210175-161210175 on Build GRCh38
rs753372460 Chr.1: 161210194-161210194 on Build GRCh38

View Full Product Details