Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 8985506-8985731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRGAP3
Entrez Gene ID
9901
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
Gene Aliases
ARHGAP14, MEGAP, SRGAP2, WRP
UniGene
Hs.654743
Gene Chromosome Location
Chr.3: 8980591-9362979 on Build GRCh38
Transcripts
NM_001033117.2 NM_014850.3 XM_011534295.1 XM_011534296.1 XM_011534301.2 XM_011534303.1 XM_017007574.1 XM_017007575.1 XM_017007576.1 XM_017007577.1 XM_017007578.1 XM_017007579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4992066 c.3263T>A p.F1088Y Chr.3: 8985556-8985556 on Build GRCh38
4120508 c.3148G>A p.A1050T Chr.3: 8985671-8985671 on Build GRCh38
4327775 c.3221C>T p.T1074I Chr.3: 8985526-8985526 on Build GRCh38
6510870 c.3204A>T p.S1068S Chr.3: 8985543-8985543 on Build GRCh38
5518403 c.3279G>A p.A1093A Chr.3: 8985540-8985540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779823946 Chr.3: 8985637-8985637 on Build GRCh38
rs1008168448 Chr.3: 8985570-8985570 on Build GRCh38
rs771190189 Chr.3: 8985709-8985709 on Build GRCh38
rs755646327 Chr.3: 8985525-8985525 on Build GRCh38
rs763590905 Chr.3: 8985657-8985657 on Build GRCh38
rs773507636 Chr.3: 8985652-8985652 on Build GRCh38
rs1055980536 Chr.3: 8985669-8985669 on Build GRCh38
rs763386146 Chr.3: 8985582-8985582 on Build GRCh38
rs376712507 Chr.3: 8985660-8985660 on Build GRCh38
rs374222241 Chr.3: 8985549-8985549 on Build GRCh38
rs765625855 Chr.3: 8985580-8985580 on Build GRCh38
rs768465759 Chr.3: 8985599-8985600 on Build GRCh38
rs769950004 Chr.3: 8985691-8985691 on Build GRCh38
rs762180219 Chr.3: 8985579-8985579 on Build GRCh38
rs766810764 Chr.3: 8985653-8985653 on Build GRCh38
rs780051953 Chr.3: 8985679-8985679 on Build GRCh38
rs146246562 Chr.3: 8985561-8985561 on Build GRCh38
rs202040708 Chr.3: 8985618-8985618 on Build GRCh38
rs61740045 Chr.3: 8985541-8985541 on Build GRCh38
rs748187826 Chr.3: 8985687-8985687 on Build GRCh38
rs764755283 Chr.3: 8985671-8985671 on Build GRCh38
rs760093750 Chr.3: 8985656-8985656 on Build GRCh38
rs758104569 Chr.3: 8985675-8985675 on Build GRCh38
rs771932247 Chr.3: 8985633-8985633 on Build GRCh38
rs919720233 Chr.3: 8985529-8985529 on Build GRCh38
rs766750795 Chr.3: 8985593-8985593 on Build GRCh38
rs779798660 Chr.3: 8985576-8985577 on Build GRCh38
rs139298704 Chr.3: 8985614-8985614 on Build GRCh38
rs112601580 Chr.3: 8985682-8985682 on Build GRCh38
rs755595138 Chr.3: 8985600-8985600 on Build GRCh38
rs770690937 Chr.3: 8985547-8985547 on Build GRCh38
rs760795714 Chr.3: 8985572-8985572 on Build GRCh38
rs538106006 Chr.3: 8985573-8985573 on Build GRCh38
rs746645536 Chr.3: 8985600-8985602 on Build GRCh38
rs535318078 Chr.3: 8985620-8985620 on Build GRCh38
rs775519092 Chr.3: 8985571-8985571 on Build GRCh38
rs773278068 Chr.3: 8985705-8985705 on Build GRCh38
rs945927419 Chr.3: 8985634-8985634 on Build GRCh38
rs200901948 Chr.3: 8985638-8985638 on Build GRCh38
rs776913830 Chr.3: 8985647-8985647 on Build GRCh38
rs750685024 Chr.3: 8985581-8985581 on Build GRCh38
rs894217708 Chr.3: 8985686-8985686 on Build GRCh38
rs1002525765 Chr.3: 8985658-8985658 on Build GRCh38
rs571850803 Chr.3: 8985642-8985642 on Build GRCh38
rs3732978 Chr.3: 8985578-8985578 on Build GRCh38
rs200111200 Chr.3: 8985680-8985680 on Build GRCh38
rs370200042 Chr.3: 8985672-8985672 on Build GRCh38
rs774821295 Chr.3: 8985655-8985655 on Build GRCh38
rs745815373 Chr.3: 8985555-8985555 on Build GRCh38
rs777317426 Chr.3: 8985606-8985606 on Build GRCh38
rs745599938 Chr.3: 8985629-8985629 on Build GRCh38
rs757842852 Chr.3: 8985574-8985575 on Build GRCh38
rs777368562 Chr.3: 8985540-8985540 on Build GRCh38
rs373709294 Chr.3: 8985641-8985641 on Build GRCh38
rs74520113 Chr.3: 8985689-8985689 on Build GRCh38
rs981733208 Chr.3: 8985659-8985659 on Build GRCh38
rs545496383 Chr.3: 8985651-8985651 on Build GRCh38
rs749577608 Chr.3: 8985708-8985708 on Build GRCh38
rs575647975 Chr.3: 8985661-8985661 on Build GRCh38
rs144096171 Chr.3: 8985597-8985597 on Build GRCh38
rs933431754 Chr.3: 8985562-8985562 on Build GRCh38

View Full Product Details