Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201478194-201478465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRADB
Entrez Gene ID
55437
Gene Name
STE20-related kinase adaptor beta
Gene Aliases
ALS2CR2, CALS-21, ILPIP, ILPIPA, PAPK, PRO1038
UniGene
Hs.652338
Gene Chromosome Location
Chr.2: 201451669-201480851 on Build GRCh38
Transcripts
NM_001206864.1 NM_018571.5 XM_005246669.1 XM_017004438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1014908 c.837G>T p.Q279H Chr.2: 201478368-201478368 on Build GRCh38
6792374 c.920G>A p.G307D Chr.2: 201478451-201478451 on Build GRCh38
6055331 c.933G>A p.G311G Chr.2: 201478464-201478464 on Build GRCh38
4510993 c.853C>T p.P285S Chr.2: 201478384-201478384 on Build GRCh38
2907286 c.875G>A p.S292N Chr.2: 201478406-201478406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931913761 Chr.2: 201478250-201478250 on Build GRCh38
rs780574357 Chr.2: 201478231-201478231 on Build GRCh38
rs375922651 Chr.2: 201478218-201478218 on Build GRCh38
rs576842105 Chr.2: 201478443-201478443 on Build GRCh38
rs11686979 Chr.2: 201478257-201478257 on Build GRCh38
rs139316950 Chr.2: 201478366-201478366 on Build GRCh38
rs745964384 Chr.2: 201478406-201478407 on Build GRCh38
rs995628093 Chr.2: 201478384-201478384 on Build GRCh38
rs779979103 Chr.2: 201478424-201478424 on Build GRCh38
rs774686337 Chr.2: 201478319-201478319 on Build GRCh38
rs900411036 Chr.2: 201478222-201478222 on Build GRCh38
rs1013267626 Chr.2: 201478430-201478430 on Build GRCh38
rs772255556 Chr.2: 201478424-201478424 on Build GRCh38
rs775113809 Chr.2: 201478357-201478357 on Build GRCh38
rs756402299 Chr.2: 201478419-201478419 on Build GRCh38
rs749446717 Chr.2: 201478435-201478435 on Build GRCh38
rs769257813 Chr.2: 201478313-201478313 on Build GRCh38
rs1015095174 Chr.2: 201478371-201478371 on Build GRCh38
rs188868611 Chr.2: 201478341-201478341 on Build GRCh38
rs764621072 Chr.2: 201478344-201478344 on Build GRCh38
rs546115190 Chr.2: 201478360-201478360 on Build GRCh38
rs756911064 Chr.2: 201478330-201478330 on Build GRCh38
rs939941512 Chr.2: 201478328-201478328 on Build GRCh38
rs368698287 Chr.2: 201478378-201478378 on Build GRCh38
rs572372910 Chr.2: 201478280-201478280 on Build GRCh38
rs762233985 Chr.2: 201478329-201478329 on Build GRCh38
rs995281706 Chr.2: 201478382-201478382 on Build GRCh38
rs143801737 Chr.2: 201478383-201478383 on Build GRCh38
rs55872461 Chr.2: 201478392-201478392 on Build GRCh38
rs11686977 Chr.2: 201478246-201478246 on Build GRCh38
rs933363608 Chr.2: 201478251-201478251 on Build GRCh38
rs755473614 Chr.2: 201478442-201478442 on Build GRCh38
rs749731340 Chr.2: 201478238-201478238 on Build GRCh38
rs907829274 Chr.2: 201478326-201478326 on Build GRCh38
rs535491501 Chr.2: 201478275-201478275 on Build GRCh38
rs769790827 Chr.2: 201478229-201478229 on Build GRCh38
rs776279874 Chr.2: 201478332-201478332 on Build GRCh38
rs757128613 Chr.2: 201478391-201478391 on Build GRCh38
rs750646416 Chr.2: 201478417-201478417 on Build GRCh38
rs868537823 Chr.2: 201478253-201478253 on Build GRCh38

View Full Product Details