Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44775995-44776258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS8
Entrez Gene ID
6202
Gene Name
ribosomal protein S8
Gene Aliases
S8
UniGene
Hs.512675
Gene Chromosome Location
Chr.1: 44775574-44778740 on Build GRCh38
Transcripts
NM_001012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1161966 c.92G>T p.R31L Chr.1: 44776121-44776121 on Build GRCh38
1162077 c.35G>A p.R12H Chr.1: 44776064-44776064 on Build GRCh38
277124 c.75G>C p.R25R Chr.1: 44776104-44776104 on Build GRCh38
6257334 c.24G>T p.W8C Chr.1: 44776053-44776053 on Build GRCh38
6769638 c.92G>A p.R31H Chr.1: 44776121-44776121 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780653740 Chr.1: 44776085-44776085 on Build GRCh38
rs994191944 Chr.1: 44776190-44776190 on Build GRCh38
rs548151280 Chr.1: 44776164-44776164 on Build GRCh38
rs368315084 Chr.1: 44776167-44776167 on Build GRCh38
rs112945716 Chr.1: 44776032-44776032 on Build GRCh38
rs540519197 Chr.1: 44776230-44776231 on Build GRCh38
rs116078889 Chr.1: 44776185-44776185 on Build GRCh38
rs1047480784 Chr.1: 44776195-44776195 on Build GRCh38
rs751388135 Chr.1: 44776170-44776170 on Build GRCh38
rs1008723599 Chr.1: 44776211-44776211 on Build GRCh38
rs374476318 Chr.1: 44776191-44776191 on Build GRCh38
rs772428183 Chr.1: 44776131-44776131 on Build GRCh38
rs762015190 Chr.1: 44776157-44776157 on Build GRCh38
rs1002100619 Chr.1: 44776212-44776212 on Build GRCh38
rs202018284 Chr.1: 44776016-44776016 on Build GRCh38
rs891241246 Chr.1: 44776196-44776196 on Build GRCh38
rs754667122 Chr.1: 44776173-44776173 on Build GRCh38
rs750477937 Chr.1: 44776165-44776165 on Build GRCh38
rs753935453 Chr.1: 44776015-44776015 on Build GRCh38
rs570070957 Chr.1: 44776202-44776202 on Build GRCh38
rs779809580 Chr.1: 44776169-44776169 on Build GRCh38
rs964212187 Chr.1: 44776228-44776228 on Build GRCh38
rs747668843 Chr.1: 44776089-44776089 on Build GRCh38
rs760170250 Chr.1: 44776113-44776113 on Build GRCh38
rs758313968 Chr.1: 44776020-44776020 on Build GRCh38
rs769497842 Chr.1: 44776092-44776092 on Build GRCh38
rs768381254 Chr.1: 44776049-44776049 on Build GRCh38
rs78902799 Chr.1: 44776219-44776219 on Build GRCh38
rs748766231 Chr.1: 44776188-44776188 on Build GRCh38
rs990400441 Chr.1: 44776069-44776069 on Build GRCh38
rs538761951 Chr.1: 44776137-44776137 on Build GRCh38
rs775954398 Chr.1: 44776133-44776133 on Build GRCh38
rs779944169 Chr.1: 44776021-44776021 on Build GRCh38
rs914789959 Chr.1: 44776080-44776080 on Build GRCh38
rs778824157 Chr.1: 44776018-44776018 on Build GRCh38
rs199571882 Chr.1: 44776187-44776187 on Build GRCh38
rs372520487 Chr.1: 44776179-44776179 on Build GRCh38
rs201664044 Chr.1: 44776062-44776062 on Build GRCh38
rs764389426 Chr.1: 44776144-44776144 on Build GRCh38
rs753990171 Chr.1: 44776146-44776146 on Build GRCh38
rs528002873 Chr.1: 44776019-44776019 on Build GRCh38
rs538975415 Chr.1: 44776204-44776204 on Build GRCh38
rs977464113 Chr.1: 44776088-44776088 on Build GRCh38
rs760897375 Chr.1: 44776134-44776134 on Build GRCh38
rs746744422 Chr.1: 44776040-44776040 on Build GRCh38
rs964324849 Chr.1: 44776048-44776048 on Build GRCh38
rs748452668 Chr.1: 44776210-44776210 on Build GRCh38
rs772837110 Chr.1: 44776109-44776109 on Build GRCh38

View Full Product Details