Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1651413-1651664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF8
Entrez Gene ID
10594
Gene Name
pre-mRNA processing factor 8
Gene Aliases
HPRP8, PRP8, PRPC8, RP13, SNRNP220
UniGene
Hs.181368
Gene Chromosome Location
Chr.17: 1650629-1684882 on Build GRCh38
Transcripts
NM_006445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718022 c.6570C>A p.P2190P Chr.17: 1651494-1651494 on Build GRCh38
5055235 c.6582C>A p.T2194T Chr.17: 1651482-1651482 on Build GRCh38
4833531 c.6507C>G p.L2169L Chr.17: 1651651-1651651 on Build GRCh38
3818871 c.6517G>A p.E2173K Chr.17: 1651547-1651547 on Build GRCh38
6847902 c.6571C>T p.Q2191* Chr.17: 1651493-1651493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779292402 Chr.17: 1651588-1651588 on Build GRCh38
rs146723312 Chr.17: 1651455-1651455 on Build GRCh38
rs779545202 Chr.17: 1651452-1651452 on Build GRCh38
rs761317928 Chr.17: 1651472-1651472 on Build GRCh38
rs199665387 Chr.17: 1651556-1651556 on Build GRCh38
rs780836561 Chr.17: 1651600-1651600 on Build GRCh38
rs1802491 Chr.17: 1651476-1651476 on Build GRCh38
rs1048004787 Chr.17: 1651551-1651551 on Build GRCh38
rs767634922 Chr.17: 1651560-1651560 on Build GRCh38
rs375103607 Chr.17: 1651604-1651604 on Build GRCh38
rs372965627 Chr.17: 1651605-1651605 on Build GRCh38
rs760803797 Chr.17: 1651629-1651629 on Build GRCh38
rs757542343 Chr.17: 1651575-1651575 on Build GRCh38
rs768288906 Chr.17: 1651597-1651597 on Build GRCh38
rs771390728 Chr.17: 1651449-1651449 on Build GRCh38
rs759259223 Chr.17: 1651611-1651611 on Build GRCh38
rs377037603 Chr.17: 1651480-1651480 on Build GRCh38
rs778417143 Chr.17: 1651571-1651571 on Build GRCh38
rs752762890 Chr.17: 1651561-1651561 on Build GRCh38
rs773192791 Chr.17: 1651479-1651479 on Build GRCh38
rs756317920 Chr.17: 1651568-1651568 on Build GRCh38
rs376347137 Chr.17: 1651603-1651603 on Build GRCh38
rs370367365 Chr.17: 1651626-1651626 on Build GRCh38
rs567754383 Chr.17: 1651616-1651616 on Build GRCh38
rs551070373 Chr.17: 1651506-1651506 on Build GRCh38
rs371715532 Chr.17: 1651440-1651440 on Build GRCh38
rs766244377 Chr.17: 1651501-1651501 on Build GRCh38
rs376002448 Chr.17: 1651466-1651466 on Build GRCh38
rs774127828 Chr.17: 1651607-1651607 on Build GRCh38
rs1007004133 Chr.17: 1651446-1651446 on Build GRCh38
rs746253603 Chr.17: 1651592-1651592 on Build GRCh38
rs929492529 Chr.17: 1651499-1651499 on Build GRCh38
rs754439139 Chr.17: 1651574-1651574 on Build GRCh38
rs368323256 Chr.17: 1651469-1651469 on Build GRCh38
rs944654762 Chr.17: 1651580-1651580 on Build GRCh38

View Full Product Details