Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10123631-10123888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRM2
Entrez Gene ID
6241
Gene Name
ribonucleotide reductase regulatory subunit M2
Gene Aliases
R2, RR2, RR2M
UniGene
Hs.226390
Gene Chromosome Location
Chr.2: 10122568-10131419 on Build GRCh38
Transcripts
NM_001034.3 NM_001165931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7144731 c.412G>A p.D138N Chr.2: 10123829-10123829 on Build GRCh38
5394011 c.537G>A p.L179L Chr.2: 10123774-10123774 on Build GRCh38
1398456 c.320T>C p.V107A Chr.2: 10123737-10123737 on Build GRCh38
5582410 c.513G>A p.K171K Chr.2: 10123750-10123750 on Build GRCh38
1237122 c.603A>G p.V201V Chr.2: 10123840-10123840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768635114 Chr.2: 10123822-10123822 on Build GRCh38
rs778499413 Chr.2: 10123725-10123725 on Build GRCh38
rs5030743 Chr.2: 10123747-10123747 on Build GRCh38
rs13400205 Chr.2: 10123737-10123737 on Build GRCh38
rs368611962 Chr.2: 10123781-10123781 on Build GRCh38
rs779484966 Chr.2: 10123757-10123757 on Build GRCh38
rs759894096 Chr.2: 10123689-10123689 on Build GRCh38
rs751351052 Chr.2: 10123722-10123722 on Build GRCh38
rs139359585 Chr.2: 10123731-10123731 on Build GRCh38
rs776760302 Chr.2: 10123835-10123835 on Build GRCh38
rs755609030 Chr.2: 10123741-10123741 on Build GRCh38
rs775749110 Chr.2: 10123694-10123694 on Build GRCh38
rs1035184008 Chr.2: 10123715-10123715 on Build GRCh38
rs373196837 Chr.2: 10123718-10123718 on Build GRCh38
rs756946380 Chr.2: 10123724-10123726 on Build GRCh38
rs746257461 Chr.2: 10123741-10123742 on Build GRCh38
rs150059735 Chr.2: 10123792-10123792 on Build GRCh38
rs72542793 Chr.2: 10123714-10123714 on Build GRCh38
rs539909033 Chr.2: 10123753-10123753 on Build GRCh38
rs750335131 Chr.2: 10123709-10123709 on Build GRCh38
rs746201114 Chr.2: 10123769-10123769 on Build GRCh38
rs764199311 Chr.2: 10123706-10123706 on Build GRCh38
rs772519994 Chr.2: 10123770-10123770 on Build GRCh38
rs147651077 Chr.2: 10123788-10123788 on Build GRCh38
rs1035175832 Chr.2: 10123733-10123733 on Build GRCh38
rs772414607 Chr.2: 10123693-10123693 on Build GRCh38
rs968501874 Chr.2: 10123778-10123778 on Build GRCh38
rs760863372 Chr.2: 10123698-10123698 on Build GRCh38
rs752296530 Chr.2: 10123738-10123738 on Build GRCh38
rs754666634 Chr.2: 10123725-10123725 on Build GRCh38
rs775888282 Chr.2: 10123780-10123780 on Build GRCh38
rs774717322 Chr.2: 10123685-10123685 on Build GRCh38
rs140269701 Chr.2: 10123754-10123754 on Build GRCh38
rs543262157 Chr.2: 10123684-10123684 on Build GRCh38

View Full Product Details