Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4946581-4946854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFN1
Entrez Gene ID
5216
Gene Name
profilin 1
Gene Aliases
ALS18
UniGene
Hs.494691
Gene Chromosome Location
Chr.17: 4945650-4949088 on Build GRCh38
Transcripts
NM_005022.3 XM_017024761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744448 c.203delG p.G68fs*6 Chr.17: 4946750-4946750 on Build GRCh38
1290721 c.167G>A p.R56Q Chr.17: 4946786-4946786 on Build GRCh38
6429329 c.146G>T p.G49V Chr.17: 4946807-4946807 on Build GRCh38
6744450 c.304A>G p.T102A Chr.17: 4946649-4946649 on Build GRCh38
5643713 c.138T>C p.A46A Chr.17: 4946815-4946815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889743070 Chr.17: 4946731-4946731 on Build GRCh38
rs764145549 Chr.17: 4946757-4946757 on Build GRCh38
rs772921326 Chr.17: 4946619-4946619 on Build GRCh38
rs387907264 Chr.17: 4946742-4946742 on Build GRCh38
rs112009460 Chr.17: 4946614-4946614 on Build GRCh38
rs760956250 Chr.17: 4946737-4946737 on Build GRCh38
rs934233454 Chr.17: 4946642-4946642 on Build GRCh38
rs372205846 Chr.17: 4946780-4946780 on Build GRCh38
rs766371231 Chr.17: 4946797-4946797 on Build GRCh38
rs879076507 Chr.17: 4946826-4946826 on Build GRCh38
rs754796691 Chr.17: 4946829-4946829 on Build GRCh38
rs747850206 Chr.17: 4946608-4946608 on Build GRCh38
rs143858310 Chr.17: 4946680-4946680 on Build GRCh38
rs749741600 Chr.17: 4946652-4946652 on Build GRCh38
rs202232499 Chr.17: 4946772-4946772 on Build GRCh38
rs751261956 Chr.17: 4946816-4946816 on Build GRCh38
rs973817714 Chr.17: 4946800-4946800 on Build GRCh38
rs774544158 Chr.17: 4946665-4946665 on Build GRCh38
rs879091547 Chr.17: 4946823-4946823 on Build GRCh38
rs915853020 Chr.17: 4946609-4946610 on Build GRCh38
rs77117613 Chr.17: 4946657-4946657 on Build GRCh38
rs11538690 Chr.17: 4946674-4946674 on Build GRCh38
rs971299035 Chr.17: 4946662-4946664 on Build GRCh38
rs1804219 Chr.17: 4946790-4946790 on Build GRCh38
rs113497606 Chr.17: 4946822-4946822 on Build GRCh38
rs372420207 Chr.17: 4946827-4946827 on Build GRCh38
rs1038366120 Chr.17: 4946828-4946828 on Build GRCh38
rs759969693 Chr.17: 4946673-4946673 on Build GRCh38
rs367732620 Chr.17: 4946721-4946721 on Build GRCh38
rs11538688 Chr.17: 4946683-4946683 on Build GRCh38
rs550290912 Chr.17: 4946653-4946653 on Build GRCh38

View Full Product Details