Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24121078-24121329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM228B
Entrez Gene ID
375190
Gene Name
family with sequence similarity 228 member B
Gene Aliases
-
UniGene
Hs.710370
Gene Chromosome Location
Chr.2: 24076874-24169638 on Build GRCh38
Transcripts
NM_001291328.1 NR_111929.1 NM_199346.2 XM_005264292.2 XM_006712007.3 XM_011532823.2

Gene Symbol
PFN4
Entrez Gene ID
375189
Gene Name
profilin family member 4
Gene Aliases
-
UniGene
Hs.442718
Gene Chromosome Location
Chr.2: 24114809-24123491 on Build GRCh38
Transcripts
NM_001291328.1 NR_111929.1 NM_199346.2 XM_005264292.2 XM_006712007.3 XM_011532823.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446610 c.137G>A p.R46Q Chr.2: 24121281-24121281 on Build GRCh38
4093161 c.122C>T p.T41M Chr.2: 24121296-24121296 on Build GRCh38
1019050 c.179G>A p.R60Q Chr.2: 24121239-24121239 on Build GRCh38
1019049 c.208G>T p.D70Y Chr.2: 24121210-24121210 on Build GRCh38
4713757 c.128G>C p.S43T Chr.2: 24121290-24121290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201033985 Chr.2: 24121303-24121303 on Build GRCh38
rs773082250 Chr.2: 24121260-24121260 on Build GRCh38
rs762616878 Chr.2: 24121306-24121306 on Build GRCh38
rs200264162 Chr.2: 24121168-24121168 on Build GRCh38
rs776497287 Chr.2: 24121137-24121137 on Build GRCh38
rs868729693 Chr.2: 24121167-24121167 on Build GRCh38
rs376361343 Chr.2: 24121282-24121282 on Build GRCh38
rs779841085 Chr.2: 24121244-24121244 on Build GRCh38
rs781053006 Chr.2: 24121241-24121241 on Build GRCh38
rs373773551 Chr.2: 24121290-24121290 on Build GRCh38
rs143462327 Chr.2: 24121239-24121239 on Build GRCh38
rs762220770 Chr.2: 24121297-24121297 on Build GRCh38
rs749978143 Chr.2: 24121205-24121205 on Build GRCh38
rs367971215 Chr.2: 24121305-24121305 on Build GRCh38
rs769661008 Chr.2: 24121154-24121154 on Build GRCh38
rs774415070 Chr.2: 24121162-24121162 on Build GRCh38
rs764889045 Chr.2: 24121266-24121266 on Build GRCh38
rs746830220 Chr.2: 24121131-24121131 on Build GRCh38
rs768465074 Chr.2: 24121136-24121136 on Build GRCh38
rs374268200 Chr.2: 24121195-24121195 on Build GRCh38
rs748102700 Chr.2: 24121140-24121140 on Build GRCh38
rs190913323 Chr.2: 24121114-24121114 on Build GRCh38
rs573212540 Chr.2: 24121194-24121194 on Build GRCh38
rs1048780716 Chr.2: 24121295-24121295 on Build GRCh38
rs762388669 Chr.2: 24121155-24121155 on Build GRCh38
rs759568336 Chr.2: 24121185-24121185 on Build GRCh38
rs188094111 Chr.2: 24121300-24121300 on Build GRCh38
rs540480895 Chr.2: 24121226-24121226 on Build GRCh38
rs772137902 Chr.2: 24121281-24121281 on Build GRCh38
rs149321469 Chr.2: 24121210-24121210 on Build GRCh38
rs779890529 Chr.2: 24121224-24121224 on Build GRCh38
rs1015577047 Chr.2: 24121252-24121252 on Build GRCh38
rs183055496 Chr.2: 24121115-24121115 on Build GRCh38
rs999139839 Chr.2: 24121151-24121151 on Build GRCh38
rs767746032 Chr.2: 24121189-24121189 on Build GRCh38
rs555639473 Chr.2: 24121231-24121231 on Build GRCh38
rs370967554 Chr.2: 24121242-24121242 on Build GRCh38
rs750205875 Chr.2: 24121187-24121187 on Build GRCh38
rs33913065 Chr.2: 24121190-24121190 on Build GRCh38
rs142176262 Chr.2: 24121202-24121202 on Build GRCh38
rs779806434 Chr.2: 24121127-24121127 on Build GRCh38
rs200145580 Chr.2: 24121296-24121296 on Build GRCh38
rs769750405 Chr.2: 24121259-24121259 on Build GRCh38
rs745993258 Chr.2: 24121265-24121265 on Build GRCh38

View Full Product Details