Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70409550-70409783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPRPN
Entrez Gene ID
58503
Gene Name
opiorphin prepropeptide
Gene Aliases
BPLP, PRL1, PROL1, opiorphin
UniGene
Hs.661425
Gene Chromosome Location
Chr.4: 70397882-70410197 on Build GRCh38
Transcripts
NM_001302807.1 NM_021225.4 NR_126503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605453 c.401A>G p.Y134C Chr.4: 70409729-70409729 on Build GRCh38
3605455 c.438C>T p.I146I Chr.4: 70409766-70409766 on Build GRCh38
1582627 c.416A>C p.N139T Chr.4: 70409744-70409744 on Build GRCh38
3605449 c.300G>A p.P100P Chr.4: 70409628-70409628 on Build GRCh38
3605451 c.373C>T p.P125S Chr.4: 70409701-70409701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773846753 Chr.4: 70409754-70409754 on Build GRCh38
rs764665833 Chr.4: 70409577-70409577 on Build GRCh38
rs752897456 Chr.4: 70409683-70409684 on Build GRCh38
rs570022371 Chr.4: 70409675-70409675 on Build GRCh38
rs763906788 Chr.4: 70409642-70409642 on Build GRCh38
rs769058859 Chr.4: 70409585-70409585 on Build GRCh38
rs751440520 Chr.4: 70409594-70409594 on Build GRCh38
rs781249696 Chr.4: 70409602-70409602 on Build GRCh38
rs756015373 Chr.4: 70409607-70409607 on Build GRCh38
rs770383722 Chr.4: 70409633-70409633 on Build GRCh38
rs781439787 Chr.4: 70409706-70409706 on Build GRCh38
rs371348643 Chr.4: 70409627-70409627 on Build GRCh38
rs865855189 Chr.4: 70409598-70409598 on Build GRCh38
rs867803285 Chr.4: 70409734-70409734 on Build GRCh38
rs763789867 Chr.4: 70409590-70409590 on Build GRCh38
rs920209067 Chr.4: 70409693-70409693 on Build GRCh38
rs762521425 Chr.4: 70409641-70409641 on Build GRCh38
rs374133764 Chr.4: 70409710-70409710 on Build GRCh38
rs746193627 Chr.4: 70409733-70409733 on Build GRCh38
rs1034248665 Chr.4: 70409717-70409717 on Build GRCh38
rs775162036 Chr.4: 70409636-70409636 on Build GRCh38
rs35096170 Chr.4: 70409573-70409573 on Build GRCh38
rs539624293 Chr.4: 70409618-70409618 on Build GRCh38
rs566298040 Chr.4: 70409606-70409606 on Build GRCh38
rs756999042 Chr.4: 70409595-70409595 on Build GRCh38
rs373411935 Chr.4: 70409632-70409632 on Build GRCh38
rs778998277 Chr.4: 70409616-70409616 on Build GRCh38
rs199955993 Chr.4: 70409575-70409575 on Build GRCh38
rs773862730 Chr.4: 70409645-70409645 on Build GRCh38
rs748866342 Chr.4: 70409750-70409750 on Build GRCh38
rs201862662 Chr.4: 70409584-70409584 on Build GRCh38
rs762126179 Chr.4: 70409662-70409662 on Build GRCh38
rs767223894 Chr.4: 70409659-70409659 on Build GRCh38
rs778024902 Chr.4: 70409680-70409680 on Build GRCh38
rs748180740 Chr.4: 70409617-70409617 on Build GRCh38
rs191965892 Chr.4: 70409752-70409752 on Build GRCh38
rs537300537 Chr.4: 70409692-70409692 on Build GRCh38
rs776137322 Chr.4: 70409736-70409736 on Build GRCh38
rs61336549 Chr.4: 70409670-70409670 on Build GRCh38
rs752809537 Chr.4: 70409671-70409671 on Build GRCh38
rs377075331 Chr.4: 70409666-70409666 on Build GRCh38
rs776994086 Chr.4: 70409630-70409630 on Build GRCh38
rs762725184 Chr.4: 70409589-70409589 on Build GRCh38
rs761473675 Chr.4: 70409658-70409658 on Build GRCh38
rs771183871 Chr.4: 70409628-70409628 on Build GRCh38

View Full Product Details