Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200887486-200887739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL3
Entrez Gene ID
53938
Gene Name
peptidylprolyl isomerase like 3
Gene Aliases
CYPJ
UniGene
Hs.121076
Gene Chromosome Location
Chr.2: 200870907-200889333 on Build GRCh38
Transcripts
NM_032472.3 NM_130906.2 XM_005246651.3 XM_005246652.4 XM_011511358.2 XM_017004352.1 XM_017004353.1 XM_017004354.1 XM_017004355.1 XM_017004356.1 XM_017004357.1 XM_017004358.1 XM_017004359.1 XM_017004360.1

SNP Detail

SNP ID Chromosome Location
rs751504561 Chr.2: 200887644-200887644 on Build GRCh38
rs756342503 Chr.2: 200887666-200887666 on Build GRCh38
rs762283963 Chr.2: 200887592-200887592 on Build GRCh38
rs571422423 Chr.2: 200887526-200887526 on Build GRCh38
rs749388531 Chr.2: 200887584-200887584 on Build GRCh38
rs750204177 Chr.2: 200887665-200887665 on Build GRCh38
rs749765329 Chr.2: 200887662-200887663 on Build GRCh38
rs764893582 Chr.2: 200887615-200887615 on Build GRCh38
rs763664463 Chr.2: 200887630-200887630 on Build GRCh38
rs978279059 Chr.2: 200887540-200887540 on Build GRCh38
rs576199938 Chr.2: 200887687-200887687 on Build GRCh38
rs557427654 Chr.2: 200887668-200887668 on Build GRCh38
rs538780774 Chr.2: 200887571-200887571 on Build GRCh38
rs767315819 Chr.2: 200887647-200887647 on Build GRCh38
rs936715366 Chr.2: 200887603-200887603 on Build GRCh38
rs537212169 Chr.2: 200887694-200887694 on Build GRCh38
rs769215833 Chr.2: 200887585-200887585 on Build GRCh38
rs757195167 Chr.2: 200887646-200887646 on Build GRCh38
rs762455164 Chr.2: 200887627-200887627 on Build GRCh38
rs374477057 Chr.2: 200887667-200887667 on Build GRCh38
rs1024761986 Chr.2: 200887522-200887522 on Build GRCh38
rs759190449 Chr.2: 200887613-200887613 on Build GRCh38
rs889886419 Chr.2: 200887648-200887648 on Build GRCh38
rs752145101 Chr.2: 200887620-200887620 on Build GRCh38
rs773488354 Chr.2: 200887602-200887602 on Build GRCh38
rs774994148 Chr.2: 200887586-200887586 on Build GRCh38
rs772457972 Chr.2: 200887600-200887600 on Build GRCh38

View Full Product Details