Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78493022-78493259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGFR
Entrez Gene ID
5737
Gene Name
prostaglandin F receptor
Gene Aliases
FP
UniGene
Hs.654365
Gene Chromosome Location
Chr.1: 78490974-78540701 on Build GRCh38
Transcripts
NM_000959.3 NM_001039585.1 XM_017001873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
95113 c.281C>T p.S94F Chr.1: 78493024-78493024 on Build GRCh38
3492574 c.427C>T p.H143Y Chr.1: 78493170-78493170 on Build GRCh38
3492570 c.365C>T p.P122L Chr.1: 78493108-78493108 on Build GRCh38
5602148 c.300C>T p.R100R Chr.1: 78493043-78493043 on Build GRCh38
6997706 c.431C>T p.S144F Chr.1: 78493174-78493174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988329081 Chr.1: 78493067-78493067 on Build GRCh38
rs780419713 Chr.1: 78493219-78493219 on Build GRCh38
rs767075974 Chr.1: 78493054-78493054 on Build GRCh38
rs754233774 Chr.1: 78493189-78493189 on Build GRCh38
rs762801817 Chr.1: 78493155-78493155 on Build GRCh38
rs774207493 Chr.1: 78493168-78493168 on Build GRCh38
rs757835420 Chr.1: 78493226-78493226 on Build GRCh38
rs772307440 Chr.1: 78493086-78493086 on Build GRCh38
rs371784163 Chr.1: 78493049-78493049 on Build GRCh38
rs375003751 Chr.1: 78493194-78493194 on Build GRCh38
rs372903767 Chr.1: 78493185-78493185 on Build GRCh38
rs755367220 Chr.1: 78493071-78493071 on Build GRCh38
rs879522256 Chr.1: 78493137-78493137 on Build GRCh38
rs200749896 Chr.1: 78493068-78493068 on Build GRCh38
rs12088246 Chr.1: 78493178-78493178 on Build GRCh38
rs766906575 Chr.1: 78493188-78493188 on Build GRCh38
rs777794939 Chr.1: 78493124-78493124 on Build GRCh38
rs779100761 Chr.1: 78493196-78493196 on Build GRCh38
rs778022964 Chr.1: 78493218-78493218 on Build GRCh38
rs905232913 Chr.1: 78493176-78493176 on Build GRCh38
rs867042175 Chr.1: 78493195-78493195 on Build GRCh38
rs749120257 Chr.1: 78493233-78493233 on Build GRCh38
rs142013001 Chr.1: 78493177-78493177 on Build GRCh38
rs76806235 Chr.1: 78493085-78493085 on Build GRCh38
rs747135810 Chr.1: 78493131-78493131 on Build GRCh38
rs745393690 Chr.1: 78493195-78493196 on Build GRCh38
rs116802591 Chr.1: 78493208-78493208 on Build GRCh38
rs769693596 Chr.1: 78493140-78493140 on Build GRCh38
rs779341659 Chr.1: 78493082-78493082 on Build GRCh38
rs146358187 Chr.1: 78493183-78493183 on Build GRCh38
rs369124989 Chr.1: 78493070-78493070 on Build GRCh38
rs1045066876 Chr.1: 78493129-78493129 on Build GRCh38
rs770979500 Chr.1: 78493227-78493227 on Build GRCh38
rs753095259 Chr.1: 78493197-78493197 on Build GRCh38
rs749974467 Chr.1: 78493059-78493059 on Build GRCh38
rs866779560 Chr.1: 78493170-78493170 on Build GRCh38
rs781334182 Chr.1: 78493231-78493231 on Build GRCh38
rs775343965 Chr.1: 78493141-78493141 on Build GRCh38
rs768266962 Chr.1: 78493160-78493160 on Build GRCh38

View Full Product Details