Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 126617155-126617416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHAX
Entrez Gene ID
51808
Gene Name
phosphorylated adaptor for RNA export
Gene Aliases
RNUXA
UniGene
Hs.555731
Gene Chromosome Location
Chr.5: 126600915-126627252 on Build GRCh38
Transcripts
NM_032177.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060215 c.842G>A p.R281Q Chr.5: 126617260-126617260 on Build GRCh38
735869 c.868C>G p.L290V Chr.5: 126617286-126617286 on Build GRCh38
6561275 c.909A>C p.Q303H Chr.5: 126617327-126617327 on Build GRCh38
7096034 c.876C>G p.L292L Chr.5: 126617294-126617294 on Build GRCh38
246559 c.848G>C p.R283T Chr.5: 126617266-126617266 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756656253 Chr.5: 126617209-126617209 on Build GRCh38
rs777368867 Chr.5: 126617328-126617328 on Build GRCh38
rs113199322 Chr.5: 126617377-126617377 on Build GRCh38
rs948896496 Chr.5: 126617307-126617307 on Build GRCh38
rs138142975 Chr.5: 126617300-126617300 on Build GRCh38
rs976062794 Chr.5: 126617201-126617201 on Build GRCh38
rs911694416 Chr.5: 126617240-126617240 on Build GRCh38
rs767933132 Chr.5: 126617288-126617288 on Build GRCh38
rs766761313 Chr.5: 126617379-126617379 on Build GRCh38
rs760060859 Chr.5: 126617385-126617385 on Build GRCh38
rs758944949 Chr.5: 126617234-126617239 on Build GRCh38
rs555987457 Chr.5: 126617346-126617346 on Build GRCh38
rs778993824 Chr.5: 126617319-126617319 on Build GRCh38
rs144262397 Chr.5: 126617331-126617331 on Build GRCh38
rs373124899 Chr.5: 126617271-126617271 on Build GRCh38
rs149600878 Chr.5: 126617302-126617302 on Build GRCh38
rs779152025 Chr.5: 126617223-126617223 on Build GRCh38
rs755176636 Chr.5: 126617216-126617216 on Build GRCh38
rs747904295 Chr.5: 126617228-126617228 on Build GRCh38
rs780679420 Chr.5: 126617246-126617246 on Build GRCh38
rs1012633282 Chr.5: 126617318-126617318 on Build GRCh38
rs766963985 Chr.5: 126617297-126617297 on Build GRCh38
rs966157266 Chr.5: 126617198-126617198 on Build GRCh38
rs370583477 Chr.5: 126617322-126617322 on Build GRCh38
rs770910416 Chr.5: 126617260-126617260 on Build GRCh38
rs377587814 Chr.5: 126617311-126617311 on Build GRCh38
rs776000263 Chr.5: 126617266-126617266 on Build GRCh38
rs775151811 Chr.5: 126617361-126617361 on Build GRCh38
rs773431331 Chr.5: 126617365-126617365 on Build GRCh38
rs373302190 Chr.5: 126617338-126617338 on Build GRCh38
rs771816965 Chr.5: 126617236-126617236 on Build GRCh38
rs775324015 Chr.5: 126617273-126617273 on Build GRCh38
rs777153407 Chr.5: 126617383-126617383 on Build GRCh38
rs777649085 Chr.5: 126617247-126617247 on Build GRCh38
rs759175447 Chr.5: 126617268-126617268 on Build GRCh38
rs761081980 Chr.5: 126617291-126617291 on Build GRCh38
rs191458650 Chr.5: 126617213-126617213 on Build GRCh38
rs769391556 Chr.5: 126617360-126617360 on Build GRCh38
rs371131850 Chr.5: 126617362-126617362 on Build GRCh38
rs745444837 Chr.5: 126617286-126617286 on Build GRCh38
rs758636231 Chr.5: 126617324-126617324 on Build GRCh38
rs571468979 Chr.5: 126617380-126617381 on Build GRCh38
rs184617288 Chr.5: 126617348-126617348 on Build GRCh38
rs746867039 Chr.5: 126617253-126617253 on Build GRCh38

View Full Product Details