Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100202257-100202496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATS
Entrez Gene ID
352954
Gene Name
GATS, stromal antigen 3 opposite strand
Gene Aliases
STAG3OS
UniGene
Hs.556063
Gene Chromosome Location
Chr.7: 100187988-100272274 on Build GRCh38
Transcripts
NM_178831.6 NR_028039.1 NR_028040.1 XM_017012154.1 XM_017012155.1 XM_017012156.1 XM_017012157.1 NM_001282716.1 NM_001282717.1 NM_001282718.1 NM_012447.3 XM_011515742.1 XM_017011683.1 XM_017011684.1 XM_017011685.1 XM_017011686.1 XM_017011687.1

Gene Symbol
STAG3
Entrez Gene ID
10734
Gene Name
stromal antigen 3
Gene Aliases
-
UniGene
Hs.592283
Gene Chromosome Location
Chr.7: 100177563-100219334 on Build GRCh38
Transcripts
NM_178831.6 NR_028039.1 NR_028040.1 XM_017012154.1 XM_017012155.1 XM_017012156.1 XM_017012157.1 NM_001282716.1 NM_001282717.1 NM_001282718.1 NM_012447.3 XM_011515742.1 XM_017011683.1 XM_017011684.1 XM_017011685.1 XM_017011686.1 XM_017011687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481363 c.2500C>T p.Q834* Chr.7: 100202277-100202277 on Build GRCh38
6548534 c.2543A>T p.Q848L Chr.7: 100202320-100202320 on Build GRCh38
1673368 c.2533G>A p.V845I Chr.7: 100202310-100202310 on Build GRCh38
6193539 c.2511A>G p.L837L Chr.7: 100202288-100202288 on Build GRCh38
5675433 c.2599C>A p.L867I Chr.7: 100202489-100202489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200352003 Chr.7: 100202461-100202461 on Build GRCh38
rs763530838 Chr.7: 100202445-100202445 on Build GRCh38
rs780730029 Chr.7: 100202316-100202316 on Build GRCh38
rs371386593 Chr.7: 100202423-100202423 on Build GRCh38
rs758794200 Chr.7: 100202388-100202388 on Build GRCh38
rs766889825 Chr.7: 100202448-100202448 on Build GRCh38
rs747261656 Chr.7: 100202429-100202429 on Build GRCh38
rs977521706 Chr.7: 100202402-100202402 on Build GRCh38
rs370339908 Chr.7: 100202312-100202312 on Build GRCh38
rs779784238 Chr.7: 100202309-100202309 on Build GRCh38
rs563157647 Chr.7: 100202392-100202392 on Build GRCh38
rs767137724 Chr.7: 100202474-100202474 on Build GRCh38
rs200369644 Chr.7: 100202324-100202324 on Build GRCh38
rs776589614 Chr.7: 100202435-100202435 on Build GRCh38
rs201079971 Chr.7: 100202298-100202298 on Build GRCh38
rs764301165 Chr.7: 100202329-100202355 on Build GRCh38
rs761074998 Chr.7: 100202372-100202372 on Build GRCh38
rs368476873 Chr.7: 100202405-100202405 on Build GRCh38
rs201540529 Chr.7: 100202443-100202443 on Build GRCh38
rs565017978 Chr.7: 100202375-100202375 on Build GRCh38
rs766679011 Chr.7: 100202413-100202413 on Build GRCh38
rs201997473 Chr.7: 100202363-100202363 on Build GRCh38
rs747677188 Chr.7: 100202408-100202408 on Build GRCh38
rs375525466 Chr.7: 100202438-100202438 on Build GRCh38
rs372921469 Chr.7: 100202323-100202323 on Build GRCh38
rs764124183 Chr.7: 100202352-100202352 on Build GRCh38
rs750338136 Chr.7: 100202366-100202367 on Build GRCh38
rs749105745 Chr.7: 100202415-100202415 on Build GRCh38
rs760933210 Chr.7: 100202345-100202345 on Build GRCh38
rs1030296788 Chr.7: 100202410-100202410 on Build GRCh38
rs775643405 Chr.7: 100202344-100202344 on Build GRCh38
rs370188308 Chr.7: 100202343-100202343 on Build GRCh38
rs768001579 Chr.7: 100202313-100202313 on Build GRCh38
rs547475730 Chr.7: 100202458-100202458 on Build GRCh38
rs766149191 Chr.7: 100202391-100202391 on Build GRCh38
rs768910328 Chr.7: 100202432-100202432 on Build GRCh38
rs773045687 Chr.7: 100202338-100202338 on Build GRCh38
rs770766325 Chr.7: 100202417-100202417 on Build GRCh38
rs754349551 Chr.7: 100202414-100202414 on Build GRCh38
rs773804705 Chr.7: 100202444-100202444 on Build GRCh38
rs879047731 Chr.7: 100202436-100202436 on Build GRCh38

View Full Product Details