Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103043207-103043438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9B2
Entrez Gene ID
133308
Gene Name
solute carrier family 9 member B2
Gene Aliases
NHA2, NHE10, NHEDC2
UniGene
Hs.546482
Gene Chromosome Location
Chr.4: 103018033-103077371 on Build GRCh38
Transcripts
NM_001300754.1 NM_001300756.1 NM_178833.5 XM_006714085.2 XM_006714086.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3660818 c.1108A>G p.M370V Chr.4: 103043334-103043334 on Build GRCh38
3940688 c.1105G>A p.V369I Chr.4: 103043337-103043337 on Build GRCh38
4121048 c.1060A>G p.S354G Chr.4: 103043382-103043382 on Build GRCh38
1426160 c.1019C>T p.T340I Chr.4: 103043423-103043423 on Build GRCh38
4002884 c.1070T>G p.F357C Chr.4: 103043372-103043372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774276171 Chr.4: 103043410-103043410 on Build GRCh38
rs776710243 Chr.4: 103043302-103043302 on Build GRCh38
rs566838288 Chr.4: 103043301-103043301 on Build GRCh38
rs774592608 Chr.4: 103043269-103043269 on Build GRCh38
rs755584478 Chr.4: 103043267-103043267 on Build GRCh38
rs774430953 Chr.4: 103043276-103043281 on Build GRCh38
rs748690773 Chr.4: 103043270-103043270 on Build GRCh38
rs770017059 Chr.4: 103043379-103043379 on Build GRCh38
rs908978663 Chr.4: 103043394-103043394 on Build GRCh38
rs746100420 Chr.4: 103043375-103043375 on Build GRCh38
rs772669553 Chr.4: 103043271-103043271 on Build GRCh38
rs757044281 Chr.4: 103043318-103043318 on Build GRCh38
rs201536768 Chr.4: 103043268-103043268 on Build GRCh38
rs367604688 Chr.4: 103043341-103043341 on Build GRCh38
rs867220327 Chr.4: 103043297-103043297 on Build GRCh38
rs750057654 Chr.4: 103043337-103043337 on Build GRCh38
rs771200383 Chr.4: 103043370-103043370 on Build GRCh38
rs201153546 Chr.4: 103043316-103043316 on Build GRCh38
rs745451805 Chr.4: 103043265-103043265 on Build GRCh38
rs2276976 Chr.4: 103043372-103043372 on Build GRCh38
rs923729808 Chr.4: 103043331-103043331 on Build GRCh38
rs150832463 Chr.4: 103043342-103043342 on Build GRCh38
rs763008517 Chr.4: 103043399-103043399 on Build GRCh38
rs868309988 Chr.4: 103043360-103043360 on Build GRCh38
rs780696639 Chr.4: 103043256-103043256 on Build GRCh38
rs762908017 Chr.4: 103043308-103043308 on Build GRCh38
rs759026079 Chr.4: 103043352-103043352 on Build GRCh38
rs747441564 Chr.4: 103043359-103043359 on Build GRCh38
rs776197546 Chr.4: 103043404-103043404 on Build GRCh38
rs767276626 Chr.4: 103043334-103043334 on Build GRCh38
rs530193538 Chr.4: 103043279-103043279 on Build GRCh38
rs538079308 Chr.4: 103043411-103043411 on Build GRCh38
rs941759306 Chr.4: 103043396-103043396 on Build GRCh38
rs1053013993 Chr.4: 103043389-103043389 on Build GRCh38
rs775586133 Chr.4: 103043383-103043383 on Build GRCh38
rs115760926 Chr.4: 103043305-103043305 on Build GRCh38
rs763987232 Chr.4: 103043312-103043312 on Build GRCh38
rs550883458 Chr.4: 103043242-103043242 on Build GRCh38
rs747353750 Chr.4: 103043294-103043294 on Build GRCh38
rs186501872 Chr.4: 103043285-103043285 on Build GRCh38
rs115461318 Chr.4: 103043353-103043353 on Build GRCh38

View Full Product Details