Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 143034054-143034325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPANXN4
Entrez Gene ID
441525
Gene Name
SPANX family member N4
Gene Aliases
CT11.9
UniGene
Hs.535082
Gene Chromosome Location
Chr.X: 143025918-143038637 on Build GRCh38
Transcripts
NM_001009613.2 XM_017029543.1 XM_017029544.1

SNP Detail

SNP ID Chromosome Location
rs200969674 Chr.X: 143034251-143034251 on Build GRCh38
rs371986808 Chr.X: 143034258-143034258 on Build GRCh38
rs781410886 Chr.X: 143034288-143034288 on Build GRCh38
rs762552303 Chr.X: 143034199-143034199 on Build GRCh38
rs752707181 Chr.X: 143034278-143034278 on Build GRCh38
rs866991217 Chr.X: 143034196-143034196 on Build GRCh38
rs771224958 Chr.X: 143034144-143034144 on Build GRCh38
rs483352701 Chr.X: 143034115-143034115 on Build GRCh38
rs757542668 Chr.X: 143034285-143034285 on Build GRCh38
rs868481032 Chr.X: 143034176-143034176 on Build GRCh38
rs922437756 Chr.X: 143034160-143034160 on Build GRCh38
rs781659042 Chr.X: 143034146-143034146 on Build GRCh38
rs768380677 Chr.X: 143034215-143034215 on Build GRCh38
rs771564677 Chr.X: 143034106-143034106 on Build GRCh38
rs202128194 Chr.X: 143034095-143034095 on Build GRCh38
rs767436379 Chr.X: 143034081-143034081 on Build GRCh38
rs748629327 Chr.X: 143034233-143034233 on Build GRCh38
rs375185866 Chr.X: 143034280-143034280 on Build GRCh38
rs185859071 Chr.X: 143034105-143034105 on Build GRCh38
rs777924370 Chr.X: 143034282-143034282 on Build GRCh38
rs865873347 Chr.X: 143034190-143034190 on Build GRCh38
rs368685767 Chr.X: 143034297-143034297 on Build GRCh38
rs10482390 Chr.X: 143034090-143034090 on Build GRCh38
rs200095139 Chr.X: 143034094-143034094 on Build GRCh38
rs779534101 Chr.X: 143034224-143034224 on Build GRCh38
rs748284794 Chr.X: 143034104-143034104 on Build GRCh38
rs550741502 Chr.X: 143034275-143034275 on Build GRCh38
rs775078391 Chr.X: 143034186-143034186 on Build GRCh38
rs770403324 Chr.X: 143034181-143034181 on Build GRCh38
rs758788318 Chr.X: 143034239-143034239 on Build GRCh38
rs777977723 Chr.X: 143034123-143034123 on Build GRCh38
rs751762205 Chr.X: 143034283-143034283 on Build GRCh38
rs893851181 Chr.X: 143034229-143034229 on Build GRCh38
rs62601929 Chr.X: 143034272-143034272 on Build GRCh38
rs568757940 Chr.X: 143034299-143034299 on Build GRCh38
rs750299685 Chr.X: 143034253-143034253 on Build GRCh38
rs746333579 Chr.X: 143034159-143034159 on Build GRCh38
rs932736751 Chr.X: 143034208-143034208 on Build GRCh38
rs574419137 Chr.X: 143034102-143034102 on Build GRCh38
rs758559375 Chr.X: 143034114-143034114 on Build GRCh38
rs780580943 Chr.X: 143034267-143034267 on Build GRCh38
rs367910737 Chr.X: 143034131-143034131 on Build GRCh38

View Full Product Details