Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20727677-20727924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI4KA
Entrez Gene ID
5297
Gene Name
phosphatidylinositol 4-kinase alpha
Gene Aliases
PI4K-ALPHA, PIK4CA, PMGYCHA, pi4K230
UniGene
Hs.529438
Gene Chromosome Location
Chr.22: 20707691-20858812 on Build GRCh38
Transcripts
NM_058004.3 XM_005261634.1 XM_005261635.1 XM_017028829.1 XM_017028830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1682121 c.4546C>T p.R1516C Chr.22: 20727827-20727827 on Build GRCh38
115711 c.4577G>A p.S1526N Chr.22: 20727796-20727796 on Build GRCh38
4102777 c.4573G>A p.V1525I Chr.22: 20727800-20727800 on Build GRCh38
4164893 c.4599+3A>G p.? Chr.22: 20727771-20727771 on Build GRCh38
4496393 c.4572C>T p.A1524A Chr.22: 20727801-20727801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866843052 Chr.22: 20727818-20727818 on Build GRCh38
rs367734763 Chr.22: 20727894-20727894 on Build GRCh38
rs767815253 Chr.22: 20727877-20727877 on Build GRCh38
rs751845006 Chr.22: 20727762-20727762 on Build GRCh38
rs764683041 Chr.22: 20727760-20727760 on Build GRCh38
rs541716120 Chr.22: 20727828-20727828 on Build GRCh38
rs112532294 Chr.22: 20727773-20727773 on Build GRCh38
rs769600910 Chr.22: 20727833-20727833 on Build GRCh38
rs772846555 Chr.22: 20727732-20727732 on Build GRCh38
rs1034836359 Chr.22: 20727886-20727886 on Build GRCh38
rs1026394802 Chr.22: 20727868-20727868 on Build GRCh38
rs575931261 Chr.22: 20727826-20727826 on Build GRCh38
rs752173158 Chr.22: 20727855-20727855 on Build GRCh38
rs371490531 Chr.22: 20727899-20727899 on Build GRCh38
rs750794403 Chr.22: 20727879-20727879 on Build GRCh38
rs750785363 Chr.22: 20727822-20727822 on Build GRCh38
rs149966923 Chr.22: 20727822-20727822 on Build GRCh38
rs150837939 Chr.22: 20727796-20727796 on Build GRCh38
rs764473705 Chr.22: 20727846-20727846 on Build GRCh38
rs772055173 Chr.22: 20727731-20727731 on Build GRCh38
rs746752211 Chr.22: 20727815-20727815 on Build GRCh38
rs200890493 Chr.22: 20727725-20727725 on Build GRCh38
rs375827091 Chr.22: 20727881-20727881 on Build GRCh38
rs777814861 Chr.22: 20727807-20727807 on Build GRCh38
rs757655457 Chr.22: 20727763-20727763 on Build GRCh38
rs762367213 Chr.22: 20727852-20727852 on Build GRCh38
rs767966254 Chr.22: 20727805-20727806 on Build GRCh38
rs763329172 Chr.22: 20727759-20727759 on Build GRCh38
rs764781504 Chr.22: 20727754-20727754 on Build GRCh38
rs34784524 Chr.22: 20727741-20727742 on Build GRCh38
rs749420571 Chr.22: 20727800-20727800 on Build GRCh38
rs181109426 Chr.22: 20727880-20727880 on Build GRCh38
rs756345848 Chr.22: 20727789-20727789 on Build GRCh38
rs1020553110 Chr.22: 20727821-20727821 on Build GRCh38
rs965954272 Chr.22: 20727782-20727782 on Build GRCh38
rs563669044 Chr.22: 20727806-20727806 on Build GRCh38
rs766721452 Chr.22: 20727830-20727838 on Build GRCh38
rs760902797 Chr.22: 20727825-20727826 on Build GRCh38
rs7289942 Chr.22: 20727745-20727745 on Build GRCh38
rs774813767 Chr.22: 20727837-20727837 on Build GRCh38
rs375002092 Chr.22: 20727900-20727900 on Build GRCh38
rs781385999 Chr.22: 20727768-20727768 on Build GRCh38
rs776465911 Chr.22: 20727747-20727747 on Build GRCh38
rs139243317 Chr.22: 20727808-20727808 on Build GRCh38
rs141464943 Chr.22: 20727794-20727794 on Build GRCh38
rs760541909 Chr.22: 20727735-20727735 on Build GRCh38
rs759114160 Chr.22: 20727748-20727748 on Build GRCh38
rs762569094 Chr.22: 20727840-20727840 on Build GRCh38
rs1007388576 Chr.22: 20727756-20727756 on Build GRCh38
rs543476122 Chr.22: 20727779-20727779 on Build GRCh38

View Full Product Details