Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230916108-230916377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC13
Entrez Gene ID
79573
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
-
UniGene
Hs.424788
Gene Chromosome Location
Chr.1: 230906241-230978872 on Build GRCh38
Transcripts
NM_001122835.2 NM_024525.4 XM_005273260.1 XM_005273261.2 XM_005273262.1 XM_005273264.2 XM_006711814.2 XM_006711815.2 XM_011544275.2 XM_011544276.2 XM_017002321.1 XM_017002322.1 XM_017002323.1 XM_017002324.1 XM_017002325.1 XM_017002326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1733954 c.2051G>A p.G684E Chr.1: 230916235-230916235 on Build GRCh38
4569794 c.2040T>C p.L680L Chr.1: 230916246-230916246 on Build GRCh38
5025536 c.2001G>A p.T667T Chr.1: 230916285-230916285 on Build GRCh38
7136091 c.2070C>A p.F690L Chr.1: 230916216-230916216 on Build GRCh38
1295987 c.2086G>A p.G696R Chr.1: 230916200-230916200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918167922 Chr.1: 230916318-230916318 on Build GRCh38
rs377084328 Chr.1: 230916207-230916207 on Build GRCh38
rs187854710 Chr.1: 230916163-230916163 on Build GRCh38
rs192530071 Chr.1: 230916184-230916184 on Build GRCh38
rs766637522 Chr.1: 230916263-230916263 on Build GRCh38
rs377754802 Chr.1: 230916334-230916334 on Build GRCh38
rs764050903 Chr.1: 230916232-230916232 on Build GRCh38
rs745577746 Chr.1: 230916223-230916223 on Build GRCh38
rs369934096 Chr.1: 230916139-230916139 on Build GRCh38
rs769711536 Chr.1: 230916333-230916333 on Build GRCh38
rs781748939 Chr.1: 230916189-230916189 on Build GRCh38
rs776597446 Chr.1: 230916237-230916237 on Build GRCh38
rs752000634 Chr.1: 230916265-230916265 on Build GRCh38
rs773885434 Chr.1: 230916208-230916208 on Build GRCh38
rs925415731 Chr.1: 230916271-230916271 on Build GRCh38
rs1029189554 Chr.1: 230916252-230916252 on Build GRCh38
rs758641365 Chr.1: 230916174-230916174 on Build GRCh38
rs753921171 Chr.1: 230916152-230916152 on Build GRCh38
rs557039950 Chr.1: 230916185-230916185 on Build GRCh38
rs576971996 Chr.1: 230916247-230916247 on Build GRCh38
rs997184959 Chr.1: 230916231-230916231 on Build GRCh38
rs369413110 Chr.1: 230916156-230916156 on Build GRCh38
rs753186650 Chr.1: 230916279-230916279 on Build GRCh38
rs1014546697 Chr.1: 230916181-230916181 on Build GRCh38
rs750501364 Chr.1: 230916261-230916261 on Build GRCh38
rs201198964 Chr.1: 230916286-230916286 on Build GRCh38
rs772862048 Chr.1: 230916143-230916143 on Build GRCh38
rs754928638 Chr.1: 230916285-230916285 on Build GRCh38
rs756644012 Chr.1: 230916281-230916281 on Build GRCh38
rs776501575 Chr.1: 230916254-230916254 on Build GRCh38
rs755343220 Chr.1: 230916269-230916269 on Build GRCh38
rs768478076 Chr.1: 230916321-230916321 on Build GRCh38
rs369821539 Chr.1: 230916229-230916229 on Build GRCh38
rs779643372 Chr.1: 230916311-230916311 on Build GRCh38
rs771744234 Chr.1: 230916224-230916224 on Build GRCh38
rs370209984 Chr.1: 230916341-230916341 on Build GRCh38
rs751590772 Chr.1: 230916327-230916327 on Build GRCh38
rs780620304 Chr.1: 230916179-230916179 on Build GRCh38
rs776481232 Chr.1: 230916326-230916326 on Build GRCh38
rs763094255 Chr.1: 230916262-230916262 on Build GRCh38
rs140000131 Chr.1: 230916270-230916270 on Build GRCh38
rs778024568 Chr.1: 230916317-230916317 on Build GRCh38
rs771718920 Chr.1: 230916302-230916302 on Build GRCh38
rs757234105 Chr.1: 230916155-230916155 on Build GRCh38
rs775277520 Chr.1: 230916228-230916228 on Build GRCh38
rs748856539 Chr.1: 230916200-230916200 on Build GRCh38
rs201962097 Chr.1: 230916197-230916197 on Build GRCh38

View Full Product Details