Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45689383-45689632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SACM1L
Entrez Gene ID
22908
Gene Name
SAC1 suppressor of actin mutations 1-like (yeast)
Gene Aliases
SAC1
UniGene
Hs.156509
Gene Chromosome Location
Chr.3: 45689241-45745425 on Build GRCh38
Transcripts
NM_001319072.1 NM_001319073.1 NM_014016.4 XM_011533500.2

SNP Detail

SNP ID Chromosome Location
rs990140775 Chr.3: 45689449-45689449 on Build GRCh38
rs775098303 Chr.3: 45689460-45689460 on Build GRCh38
rs774015042 Chr.3: 45689434-45689434 on Build GRCh38
rs188809579 Chr.3: 45689590-45689590 on Build GRCh38
rs535347679 Chr.3: 45689518-45689518 on Build GRCh38
rs748917866 Chr.3: 45689520-45689520 on Build GRCh38
rs887406767 Chr.3: 45689602-45689602 on Build GRCh38
rs557264197 Chr.3: 45689585-45689585 on Build GRCh38
rs183358538 Chr.3: 45689445-45689445 on Build GRCh38
rs200675699 Chr.3: 45689453-45689453 on Build GRCh38
rs923447460 Chr.3: 45689483-45689483 on Build GRCh38
rs763837572 Chr.3: 45689481-45689481 on Build GRCh38
rs777456932 Chr.3: 45689425-45689425 on Build GRCh38
rs1048974860 Chr.3: 45689591-45689591 on Build GRCh38
rs897690514 Chr.3: 45689584-45689584 on Build GRCh38
rs748572726 Chr.3: 45689415-45689415 on Build GRCh38
rs369162043 Chr.3: 45689531-45689531 on Build GRCh38
rs760498810 Chr.3: 45689474-45689474 on Build GRCh38
rs1040106879 Chr.3: 45689454-45689454 on Build GRCh38
rs1037999176 Chr.3: 45689513-45689513 on Build GRCh38
rs1012596752 Chr.3: 45689608-45689608 on Build GRCh38
rs750652021 Chr.3: 45689482-45689482 on Build GRCh38
rs933480790 Chr.3: 45689488-45689488 on Build GRCh38
rs778527414 Chr.3: 45689534-45689534 on Build GRCh38
rs771759230 Chr.3: 45689542-45689542 on Build GRCh38
rs756467545 Chr.3: 45689512-45689512 on Build GRCh38
rs1025883044 Chr.3: 45689409-45689409 on Build GRCh38
rs1050639432 Chr.3: 45689489-45689489 on Build GRCh38
rs929704770 Chr.3: 45689569-45689569 on Build GRCh38
rs745393331 Chr.3: 45689540-45689540 on Build GRCh38
rs907030062 Chr.3: 45689594-45689594 on Build GRCh38
rs748964004 Chr.3: 45689426-45689426 on Build GRCh38
rs570535617 Chr.3: 45689427-45689427 on Build GRCh38
rs769279093 Chr.3: 45689418-45689418 on Build GRCh38
rs899833371 Chr.3: 45689547-45689547 on Build GRCh38
rs148929521 Chr.3: 45689490-45689490 on Build GRCh38
rs866989771 Chr.3: 45689484-45689484 on Build GRCh38
rs1034226888 Chr.3: 45689429-45689429 on Build GRCh38
rs969931921 Chr.3: 45689433-45689433 on Build GRCh38
rs755434397 Chr.3: 45689511-45689511 on Build GRCh38
rs914698320 Chr.3: 45689452-45689452 on Build GRCh38
rs1040213992 Chr.3: 45689574-45689574 on Build GRCh38
rs766388124 Chr.3: 45689504-45689504 on Build GRCh38
rs751641902 Chr.3: 45689508-45689508 on Build GRCh38

View Full Product Details