Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 33379797-33380022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT10
Entrez Gene ID
341359
Gene Name
synaptotagmin 10
Gene Aliases
-
UniGene
Hs.118703
Gene Chromosome Location
Chr.12: 33375413-33439819 on Build GRCh38
Transcripts
NM_198992.3 XM_011520644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072437 c.1500G>C p.E500D Chr.12: 33379832-33379832 on Build GRCh38
5665680 c.1489C>T p.P497S Chr.12: 33379843-33379843 on Build GRCh38
4778925 c.1430G>T p.G477V Chr.12: 33379902-33379902 on Build GRCh38
6072436 c.1432C>A p.R478R Chr.12: 33379900-33379900 on Build GRCh38
4778922 c.1431G>T p.G477G Chr.12: 33379901-33379901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753276345 Chr.12: 33379939-33379939 on Build GRCh38
rs779412084 Chr.12: 33379867-33379867 on Build GRCh38
rs574136680 Chr.12: 33379910-33379910 on Build GRCh38
rs138986207 Chr.12: 33379853-33379853 on Build GRCh38
rs771335075 Chr.12: 33379992-33379992 on Build GRCh38
rs34190017 Chr.12: 33379869-33379869 on Build GRCh38
rs750428498 Chr.12: 33379923-33379923 on Build GRCh38
rs758904405 Chr.12: 33379935-33379935 on Build GRCh38
rs758783637 Chr.12: 33379976-33379976 on Build GRCh38
rs759867053 Chr.12: 33379899-33379899 on Build GRCh38
rs760501861 Chr.12: 33379926-33379926 on Build GRCh38
rs543371391 Chr.12: 33379993-33379993 on Build GRCh38
rs778542237 Chr.12: 33379964-33379964 on Build GRCh38
rs866542712 Chr.12: 33379938-33379938 on Build GRCh38
rs142586141 Chr.12: 33379875-33379875 on Build GRCh38
rs765981943 Chr.12: 33379929-33379929 on Build GRCh38
rs770125863 Chr.12: 33379900-33379900 on Build GRCh38
rs776540776 Chr.12: 33379889-33379889 on Build GRCh38
rs752304716 Chr.12: 33379969-33379969 on Build GRCh38
rs747589678 Chr.12: 33379986-33379986 on Build GRCh38
rs754427228 Chr.12: 33379956-33379956 on Build GRCh38
rs150924840 Chr.12: 33379850-33379850 on Build GRCh38
rs753400818 Chr.12: 33379872-33379872 on Build GRCh38
rs755755759 Chr.12: 33379858-33379858 on Build GRCh38
rs776932613 Chr.12: 33379832-33379832 on Build GRCh38
rs767376234 Chr.12: 33379916-33379916 on Build GRCh38
rs376432626 Chr.12: 33379912-33379912 on Build GRCh38
rs866874853 Chr.12: 33379885-33379885 on Build GRCh38
rs777975949 Chr.12: 33379982-33379982 on Build GRCh38
rs369419995 Chr.12: 33379963-33379963 on Build GRCh38
rs150486538 Chr.12: 33379859-33379859 on Build GRCh38
rs771090310 Chr.12: 33379886-33379886 on Build GRCh38
rs778375475 Chr.12: 33379877-33379877 on Build GRCh38
rs748526182 Chr.12: 33379873-33379873 on Build GRCh38
rs755675734 Chr.12: 33379866-33379866 on Build GRCh38
rs747425032 Chr.12: 33379879-33379879 on Build GRCh38
rs545012156 Chr.12: 33379854-33379854 on Build GRCh38

View Full Product Details