Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38223745-38223980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXSR1
Entrez Gene ID
9943
Gene Name
oxidative stress responsive 1
Gene Aliases
OSR1
UniGene
Hs.475970
Gene Chromosome Location
Chr.3: 38165051-38255488 on Build GRCh38
Transcripts
NM_005109.2 XM_005265638.2 XM_011534331.2 XM_017007601.1 XM_017007602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6010921 c.669G>A p.G223G Chr.3: 38223880-38223880 on Build GRCh38
5630568 c.619A>C p.K207Q Chr.3: 38223830-38223830 on Build GRCh38
5899945 c.677C>T p.P226L Chr.3: 38223888-38223888 on Build GRCh38
7295756 c.663T>C p.A221A Chr.3: 38223874-38223874 on Build GRCh38
6846922 c.682C>T p.H228Y Chr.3: 38223893-38223893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768642265 Chr.3: 38223883-38223883 on Build GRCh38
rs202064487 Chr.3: 38223790-38223790 on Build GRCh38
rs774133588 Chr.3: 38223920-38223920 on Build GRCh38
rs913448864 Chr.3: 38223881-38223881 on Build GRCh38
rs778152730 Chr.3: 38223781-38223781 on Build GRCh38
rs766994566 Chr.3: 38223934-38223934 on Build GRCh38
rs148501279 Chr.3: 38223815-38223815 on Build GRCh38
rs1051174393 Chr.3: 38223826-38223826 on Build GRCh38
rs772139791 Chr.3: 38223836-38223836 on Build GRCh38
rs754035874 Chr.3: 38223799-38223799 on Build GRCh38
rs770757592 Chr.3: 38223918-38223918 on Build GRCh38
rs922089142 Chr.3: 38223775-38223775 on Build GRCh38
rs756449681 Chr.3: 38223770-38223770 on Build GRCh38
rs866030348 Chr.3: 38223811-38223811 on Build GRCh38
rs934019667 Chr.3: 38223824-38223824 on Build GRCh38
rs889896943 Chr.3: 38223862-38223862 on Build GRCh38
rs762812182 Chr.3: 38223882-38223882 on Build GRCh38
rs753165849 Chr.3: 38223955-38223955 on Build GRCh38
rs746947946 Chr.3: 38223798-38223798 on Build GRCh38
rs370524330 Chr.3: 38223925-38223925 on Build GRCh38
rs746001398 Chr.3: 38223825-38223825 on Build GRCh38
rs773022296 Chr.3: 38223876-38223876 on Build GRCh38
rs769501705 Chr.3: 38223874-38223874 on Build GRCh38
rs748063783 Chr.3: 38223869-38223869 on Build GRCh38
rs151105173 Chr.3: 38223817-38223817 on Build GRCh38
rs145105894 Chr.3: 38223863-38223863 on Build GRCh38

View Full Product Details