Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44522070-44522325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPEAR
Entrez Gene ID
54084
Gene Name
thrombospondin type laminin G domain and EAR repeats
Gene Aliases
C21orf29, DFNB98, TSP-EAR
UniGene
Hs.660703
Gene Chromosome Location
Chr.21: 44497892-44711580 on Build GRCh38
Transcripts
NM_001272037.1 NM_144991.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1685787 c.1356_1357delCA p.D452fs*105 Chr.21: 44522092-44522093 on Build GRCh38
7348420 c.1339G>A p.D447N Chr.21: 44522110-44522110 on Build GRCh38
1031295 c.1353C>T p.I451I Chr.21: 44522096-44522096 on Build GRCh38
6279267 c.1379C>T p.P460L Chr.21: 44522070-44522070 on Build GRCh38
6094867 c.1359T>G p.S453R Chr.21: 44522090-44522090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781806143 Chr.21: 44522099-44522099 on Build GRCh38
rs927177781 Chr.21: 44522245-44522245 on Build GRCh38
rs949831951 Chr.21: 44522110-44522110 on Build GRCh38
rs587596608 Chr.21: 44522176-44522176 on Build GRCh38
rs889523289 Chr.21: 44522204-44522204 on Build GRCh38
rs112588586 Chr.21: 44522232-44522232 on Build GRCh38
rs143347822 Chr.21: 44522105-44522106 on Build GRCh38
rs233230 Chr.21: 44522228-44522228 on Build GRCh38
rs76470396 Chr.21: 44522272-44522272 on Build GRCh38
rs587638231 Chr.21: 44522305-44522305 on Build GRCh38
rs782525274 Chr.21: 44522119-44522119 on Build GRCh38
rs189461048 Chr.21: 44522207-44522207 on Build GRCh38
rs587756873 Chr.21: 44522298-44522298 on Build GRCh38
rs587745506 Chr.21: 44522227-44522227 on Build GRCh38
rs901431000 Chr.21: 44522303-44522303 on Build GRCh38
rs782596331 Chr.21: 44522128-44522128 on Build GRCh38
rs371240458 Chr.21: 44522137-44522137 on Build GRCh38
rs587634035 Chr.21: 44522223-44522223 on Build GRCh38
rs901439518 Chr.21: 44522174-44522174 on Build GRCh38
rs984199299 Chr.21: 44522267-44522267 on Build GRCh38
rs782052734 Chr.21: 44522150-44522150 on Build GRCh38
rs115979275 Chr.21: 44522264-44522264 on Build GRCh38
rs144318001 Chr.21: 44522214-44522214 on Build GRCh38
rs587606267 Chr.21: 44522134-44522134 on Build GRCh38
rs369718755 Chr.21: 44522148-44522148 on Build GRCh38
rs147568199 Chr.21: 44522138-44522138 on Build GRCh38
rs386819161 Chr.21: 44522227-44522229 on Build GRCh38
rs782405411 Chr.21: 44522154-44522154 on Build GRCh38
rs148471285 Chr.21: 44522111-44522111 on Build GRCh38
rs1036995329 Chr.21: 44522297-44522297 on Build GRCh38
rs917186860 Chr.21: 44522095-44522095 on Build GRCh38
rs1037215861 Chr.21: 44522167-44522167 on Build GRCh38
rs587652857 Chr.21: 44522096-44522096 on Build GRCh38
rs940044694 Chr.21: 44522163-44522163 on Build GRCh38
rs782647345 Chr.21: 44522140-44522140 on Build GRCh38
rs781967339 Chr.21: 44522149-44522149 on Build GRCh38
rs781849166 Chr.21: 44522115-44522115 on Build GRCh38
rs888478656 Chr.21: 44522160-44522160 on Build GRCh38
rs587755630 Chr.21: 44522240-44522240 on Build GRCh38
rs939989330 Chr.21: 44522273-44522273 on Build GRCh38
rs202064846 Chr.21: 44522093-44522093 on Build GRCh38
rs781832854 Chr.21: 44522131-44522131 on Build GRCh38
rs62218855 Chr.21: 44522143-44522143 on Build GRCh38
rs587679999 Chr.21: 44522212-44522212 on Build GRCh38
rs782352710 Chr.21: 44522129-44522129 on Build GRCh38
rs78026972 Chr.21: 44522224-44522224 on Build GRCh38
rs587748102 Chr.21: 44522205-44522205 on Build GRCh38
rs782592457 Chr.21: 44522136-44522136 on Build GRCh38
rs73377696 Chr.21: 44522177-44522177 on Build GRCh38

View Full Product Details