Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203496908-203497179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPTC
Entrez Gene ID
26254
Gene Name
opticin
Gene Aliases
OPT
UniGene
Hs.632468
Gene Chromosome Location
Chr.1: 203494143-203508949 on Build GRCh38
Transcripts
NM_014359.3 XM_011509406.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
678062 c.269G>T p.S90I Chr.1: 203497014-203497014 on Build GRCh38
5550731 c.274G>A p.A92T Chr.1: 203497019-203497019 on Build GRCh38
7329545 c.249C>A p.L83L Chr.1: 203496994-203496994 on Build GRCh38
4537189 c.238G>A p.V80M Chr.1: 203496983-203496983 on Build GRCh38
1748089 c.294A>T p.P98P Chr.1: 203497039-203497039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114392920 Chr.1: 203497033-203497033 on Build GRCh38
rs374413510 Chr.1: 203496968-203496968 on Build GRCh38
rs775246938 Chr.1: 203497144-203497144 on Build GRCh38
rs753952930 Chr.1: 203497072-203497072 on Build GRCh38
rs778306661 Chr.1: 203497107-203497107 on Build GRCh38
rs756225137 Chr.1: 203497011-203497011 on Build GRCh38
rs766101838 Chr.1: 203496955-203496955 on Build GRCh38
rs143593924 Chr.1: 203497116-203497116 on Build GRCh38
rs528474569 Chr.1: 203496953-203496953 on Build GRCh38
rs780224108 Chr.1: 203497012-203497012 on Build GRCh38
rs922169346 Chr.1: 203497031-203497031 on Build GRCh38
rs997537054 Chr.1: 203497115-203497115 on Build GRCh38
rs771569384 Chr.1: 203497126-203497126 on Build GRCh38
rs764902482 Chr.1: 203496984-203496984 on Build GRCh38
rs779076203 Chr.1: 203497032-203497032 on Build GRCh38
rs139974902 Chr.1: 203497051-203497051 on Build GRCh38
rs778467956 Chr.1: 203497034-203497034 on Build GRCh38
rs772651433 Chr.1: 203496951-203496951 on Build GRCh38
rs776915483 Chr.1: 203496995-203496995 on Build GRCh38
rs868156612 Chr.1: 203497112-203497112 on Build GRCh38
rs761757677 Chr.1: 203497066-203497066 on Build GRCh38
rs775617361 Chr.1: 203497074-203497074 on Build GRCh38
rs779265728 Chr.1: 203497079-203497079 on Build GRCh38
rs199853181 Chr.1: 203497068-203497068 on Build GRCh38
rs144226194 Chr.1: 203497137-203497137 on Build GRCh38
rs199927981 Chr.1: 203497101-203497101 on Build GRCh38
rs907985750 Chr.1: 203496994-203496994 on Build GRCh38
rs755255988 Chr.1: 203497028-203497028 on Build GRCh38
rs1056738509 Chr.1: 203497151-203497151 on Build GRCh38
rs764075452 Chr.1: 203497006-203497006 on Build GRCh38
rs750496460 Chr.1: 203497010-203497010 on Build GRCh38
rs200144282 Chr.1: 203496952-203496952 on Build GRCh38
rs372256499 Chr.1: 203497103-203497103 on Build GRCh38
rs774797311 Chr.1: 203496934-203496934 on Build GRCh38
rs61731863 Chr.1: 203497062-203497062 on Build GRCh38
rs776313749 Chr.1: 203497046-203497046 on Build GRCh38
rs146439963 Chr.1: 203497036-203497036 on Build GRCh38
rs768388380 Chr.1: 203497146-203497146 on Build GRCh38
rs149657301 Chr.1: 203497056-203497056 on Build GRCh38
rs752541264 Chr.1: 203496991-203496991 on Build GRCh38
rs114751507 Chr.1: 203497050-203497050 on Build GRCh38
rs771360927 Chr.1: 203496977-203496977 on Build GRCh38
rs368732612 Chr.1: 203497058-203497058 on Build GRCh38
rs990831793 Chr.1: 203496957-203496957 on Build GRCh38
rs370126695 Chr.1: 203496965-203496965 on Build GRCh38
rs983525834 Chr.1: 203496990-203496990 on Build GRCh38
rs375900748 Chr.1: 203496980-203496980 on Build GRCh38
rs567505517 Chr.1: 203497016-203497016 on Build GRCh38
rs751545013 Chr.1: 203497063-203497063 on Build GRCh38
rs28462337 Chr.1: 203497138-203497138 on Build GRCh38
rs370187993 Chr.1: 203497145-203497145 on Build GRCh38
rs186768430 Chr.1: 203496947-203496947 on Build GRCh38
rs774159572 Chr.1: 203497147-203497147 on Build GRCh38
rs536165537 Chr.1: 203497018-203497018 on Build GRCh38

View Full Product Details